Skip to Content
Merck
All Photos(3)

Key Documents

185701

Sigma-Aldrich

Benzylamine

ReagentPlus®, 99%

Synonym(s):

α-Aminotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NH2
CAS Number:
Molecular Weight:
107.15
Beilstein:
741984
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

99%

form

liquid

refractive index

n20/D 1.543 (lit.)

bp

184-185 °C (lit.)

mp

10 °C (lit.)

solubility

alcohol: miscible
diethyl ether: miscible
water: miscible

density

0.981 g/mL at 25 °C (lit.)

SMILES string

NCc1ccccc1

InChI

1S/C7H9N/c8-6-7-4-2-1-3-5-7/h1-5H,6,8H2

InChI key

WGQKYBSKWIADBV-UHFFFAOYSA-N

Gene Information

human ... AOC3(8639)
mouse ... Aoc3(11754)

Looking for similar products? Visit Product Comparison Guide

General description

Benzylamine (BnNH2) is an aromatic amine widely used for the production of organic fine chemicals and many pharmaceutical compounds. It can be prepared by the hydrogenation of benzonitrile and reductive amination of benzaldehyde using a metal catalyst.
Benzylamine undergoes oxidative deamination reaction catalyzed by benzylamine oxidase and monoamine oxidase type B.

Application

  • Benzylamine was used in modification of alkanethiol monolayers via amide and ester formation reactions for attachment onto metal electrodes.
  • It was used as carrier electrolyte for separation of alkali, alkaline earth and ammonium cations in water samples by capillary electrophoresis with indirect UV detection.
  • It was used in synthesis of cross-linked porous copolymer supports based on N-(p-vinylbenzoyl)-2-methylalanine, styrene and divinylbenzene.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aida Mohammadi et al.
Sensors (Basel, Switzerland), 18(10) (2018-10-14)
This study examines the improvements in surface-enhanced Raman scattering (SERS) performance achieved when silver nanodendritic structures are coated with various graphene-based materials, namely graphene oxide (GO), reduced graphene oxide (rGO), and graphene nanoplatelets (GNPs). The tests are performed on our
Benzylamine
Heuer, Lutz
Ullmann's Encyclopedia of Industrial Chemistry (2000)
Hydrogenation of benzonitrile to benzylamine catalyzed by ruthenium hydride complexes with P- NH- NH- P tetradentate ligands: evidence for a hydridic- protonic outer sphere mechanism
Li Tianshu, et al.
Organometallics, 26(24), 5940-5949 (2007)
Capillary electrophoresis of alkali and alkaline-earth cations with imidazole or benzylamine buffers.
Morin PCF and Dreux M.
Journal of Liquid Chromatography and Related Technologies, 17(18), 3869-3888 (1994)
Preparation and characterization of azlactone functionalized polymer supports and their application as scavengers.
Guyomard A, et al.
Eur. Polymer J., 40(10), 2343-2348 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service