Skip to Content
Merck
All Photos(1)

Key Documents

46209

Supelco

Picric acid solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Synonym(s):

2,4,6-Trinitrophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)3C6H2OH
CAS Number:
Molecular Weight:
229.10
Beilstein:
423400
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

Oc1c(cc(cc1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H3N3O7/c10-6-4(8(13)14)1-3(7(11)12)2-5(6)9(15)16/h1-2,10H

InChI key

OXNIZHLAWKMVMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aiyun Li et al.
EBioMedicine, 34, 243-255 (2018-08-11)
Mitochondrial pyruvate import via mitochondrial pyruvate carrier (MPC) is a central step in hepatic gluconeogenesis. Berberine inhibits hepatic gluconeogenesis, but the mechanism is incompletely understood. This study aims to investigate whether berberine could reduce excessive hepatic glucose production (HGP) by
Michael K Appenteng et al.
Molecules (Basel, Switzerland), 26(5) (2021-04-04)
Cyanogenic glycosides (CNGs) are naturally occurring plant molecules (nitrogenous plant secondary metabolites) which consist of an aglycone and a sugar moiety. Hydrogen cyanide (HCN) is released from these compounds following enzymatic hydrolysis causing potential toxicity issues. The presence of CNGs
Debby Kruijsen et al.
Journal of virology, 87(13), 7550-7557 (2013-05-03)
Infants are protected from a severe respiratory syncytial virus (RSV) infection in the first months of life by maternal antibodies or by prophylactically administered neutralizing antibodies. Efforts are under way to produce RSV-specific antibodies with increased neutralizing capacity compared to
Zhonghua Xiang et al.
Macromolecular rapid communications, 33(14), 1184-1190 (2012-04-18)
Three porous luminescent covalent--organic polymers (COPs) have been synthesized through self-polycondensation of the monomers of tris(4-bromophenyl)amine, 1,3,5-tris(4-bromophenyl)benzene, and 2,4,6-tris-(4-bromo-phenyl)-[1,3,5]triazine by using Ni-catalyzed Yamamoto reaction. All the COP materials possess not only high Brunauer-Emmett-Teller (BET) specific surface area of about 2000
Giuliana Magri et al.
Nature immunology, 15(4), 354-364 (2014-02-25)
Innate lymphoid cells (ILCs) regulate stromal cells, epithelial cells and cells of the immune system, but their effect on B cells remains unclear. Here we identified RORγt(+) ILCs near the marginal zone (MZ), a splenic compartment that contains innate-like B

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service