Skip to Content
Merck
All Photos(1)

Key Documents

46018

Supelco

Nonylphenol

PESTANAL®, analytical standard, technical mixture

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
Beilstein:
2047450
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.511 (lit.)

density

0.937 g/mL at 25 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCCc1ccc(O)cc1

InChI

1S/C15H24O/c1-13(2)7-5-3-4-6-8-14-9-11-15(16)12-10-14/h9-13,16H,3-8H2,1-2H3

InChI key

JSFITYFUKSFPBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonylphenol belongs to the class of alkylphenols, commonly employed as a precursor for the commercially important nonylphenol polyethoxylate, a non-ionic surfactant that is widely used for a broad-spectrum of applications in commercial and household formulations.

Application

Nonylphenol may be used as an analytical reference standard for the quantification of the analyte in environmental samples using various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-phase extraction of bisphenol A, nonylphenol and 4-octylphenol from environmental water samples using microporous bamboo charcoal, and their determination by HPLC
Zhao S-R, et al.
Microchimica Acta, 165(3-4), 443-447 (2009)
Maurizio Forte et al.
Ecotoxicology and environmental safety, 180, 412-419 (2019-05-21)
Estrogens play a role in the patho-physiology of the prostate. In the present work we studied the effects of nonylphenol (NP), a xenoestrogen, on human adenocarcinoma prostate cells (LNCaP). In order to understand molecular and cellular involvement, we observed the
Simultaneous determination of the endocrine disrupting compounds nonylphenol, nonylphenol ethoxylates, triclosan and bisphenol A in wastewater and sewage sludge by gas chromatography-mass spectrometry.
Gatidou G, et al.
Journal of Chromatography A, 1138(1-2), 32-41 (2007)
Evangelia Terzopoulou et al.
Environmental science and pollution research international, 22(2), 1095-1112 (2014-08-12)
A multi-residue method, based on gas chromatography coupled to tandem mass spectrometry (GC-MS/MS), has been developed for the determination of 70 organic micropollutants from various chemical classes (organochlorinated, organophosphorous, triazines, carbamate and urea, polycyclic aromatic hydrocarbons, polychlorinated biphenyls, pharmaceuticals, phenols
Fengxia Wu et al.
Aquatic toxicology (Amsterdam, Netherlands), 155, 43-51 (2014-06-30)
We studied the effects of endocrine disrupting compounds nonylphenol (NP) and letrozole (LE) on the male goldfish Carassius aumtus. Exposure to NP (20 μg l(-1)) alone caused a significant up-regulation in the expression of aromatase, estrogen receptors and vitellogenin (VTG)

Articles

Nonylphenols (NP) are ubiquitous substances that have been detected in highly diverse foodstuff. As breakdown products of nonylphenol ethoxylates, NP could end up in the aquatic environment. This article presents a rapid method for NP detection in surface water by planar solid phase extraction – planar yeast estrogen screen (pSPE-pYES).

Nonylphenols (NP) are ubiquitous substances that have been detected in highly diverse foodstuff. As breakdown products of nonylphenol ethoxylates, NP could end up in the aquatic environment. This article presents a rapid method for NP detection in surface water by planar solid phase extraction – planar yeast estrogen screen (pSPE-pYES).

Nonylphenols (NP) are ubiquitous substances that have been detected in highly diverse foodstuff. As breakdown products of nonylphenol ethoxylates, NP could end up in the aquatic environment. This article presents a rapid method for NP detection in surface water by planar solid phase extraction – planar yeast estrogen screen (pSPE-pYES).

Nonylphenols (NP) are ubiquitous substances that have been detected in highly diverse foodstuff. As breakdown products of nonylphenol ethoxylates, NP could end up in the aquatic environment. This article presents a rapid method for NP detection in surface water by planar solid phase extraction – planar yeast estrogen screen (pSPE-pYES).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service