Skip to Content
Merck
All Photos(1)

Key Documents

45653

Supelco

Quintozene

PESTANAL®, analytical standard

Synonym(s):

Pentachloronitrobenzene, PCNB, Quintozene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Cl5NO2
CAS Number:
Molecular Weight:
295.33
Beilstein:
1914324
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

140-143 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

[O-][N+](=O)c1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl

InChI

1S/C6Cl5NO2/c7-1-2(8)4(10)6(12(13)14)5(11)3(1)9

InChI key

LKPLKUMXSAEKID-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quintozene is a pentachloronitrobenzene (PCNB) fungicide that is primarily used to control “snow mold” on golf courses.

Application

Quintozene may be used as an internal standard for the determination of endocrine disrupter compounds (EDCs) in fish using modified QuEChERS and gas chromatography coupled with mass spectrometry in tandem (GC–MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a multiresidue method for the determination of endocrine disrupters in fish fillet using gas chromatography--triple quadrupole tandem mass spectrometry.
Munaretto J S, et al.
Talanta, 116, 827-834 (2013)
Ying Ying Li et al.
Journal of environmental management, 119, 143-150 (2013-03-12)
Pentachloronitrobenzene (PCNB) is a fungicide belonging to the organochlorine family and used extensively in agriculture for crop production. Many studies have implied that PCNB has become an environmental concern due to its widespread contamination in eco-systems. However, whether PCNB is
Didem Okutman Tas et al.
Journal of agricultural and food chemistry, 55(14), 5390-5398 (2007-06-08)
The effect of pH and temperature on the microbial reductive transformation of pentachloronitrobenzene (PCNB), an organochlorine fungicide, was investigated with a mixed fermentative/methanogenic culture developed from a contaminated estuarine sediment. Culture series were incubated at a temperature range from 4
Weiguo Zhang et al.
Rapid communications in mass spectrometry : RCM, 20(10), 1563-1568 (2006-04-22)
The effects of overlapping levels and concentration ratios of overlapping components, and of scan rates of the mass spectrometer, on the capability of the automated mass spectral deconvolution and identification system (AMDIS) in pesticide residue analysis were studied. To investigate
Wang Zhang et al.
Journal of hazardous materials, 198, 275-281 (2011-11-15)
Pentachloronitrobenzene (PCNB) has been identified as a potential persistent organic pollutant, and received concerns from many countries for regulation due to its impurities such as dioxins and furans. Considering the need for its disposal, the mechanochemical destruction approach was investigated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service