Skip to Content
Merck
All Photos(1)

Key Documents

45460

Supelco

Disulfoton

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H19O2PS3
CAS Number:
Molecular Weight:
274.40
Beilstein:
1709167
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)SCCSCC

InChI

1S/C8H19O2PS3/c1-4-9-11(12,10-5-2)14-8-7-13-6-3/h4-8H2,1-3H3

InChI key

DOFZAZXDOSGAJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

271.4 °F

Flash Point(C)

133 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T A Johnson et al.
Journal of economic entomology, 100(4), 1328-1334 (2007-09-14)
The linden borer, Saperda vestita Say (Coleoptera: Cerambycidae), is a native insect species that is common throughout north central and northeastern North America. Over the past decade, increasing occurrence of damage associated with the linden borer has been reported on
T Zahn et al.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie, 48(1), 47-64 (1996-01-01)
Ultrastructural, stereological and biochemical alterations in isolated hepatocytes and the permanent fibrocyte-like cell line R1 from rainbow trout (Oncorhynchus mykiss) exposed to 0, 0.2, 2 and 20 mg/l of the phosphorodithioate pesticide disulfoton (Solvirex, O,O-diethyl S-2-ethylthioethyl phosphorodithioate) for up to
B diSioudi et al.
Biochemistry, 38(10), 2866-2872 (1999-03-13)
Organophosphorus hydrolase (OPH, EC 8.1.3.1) is a dimeric, bacterial enzyme that detoxifies many organophosphorus neurotoxins by hydrolyzing a variety of phosphonate bonds. The histidinyl residues at amino acid positions 254 and 257 are located near the bimetallic active site present
Qiu Gan et al.
Journal of agricultural and food chemistry, 54(20), 7753-7760 (2006-09-28)
The reactions of thiometon and its ethyl analogue, disulfoton, with reduced sulfur species [e.g., bisulfide (HS-), polysulfide (S(n)2-), thiophenolate (PhS-), and thiosulfate (S2O3(2-))] were examined in well-defined aqueous solutions under anoxic conditions. The role of reduced sulfur species was investigated
H Arnold et al.
Ecotoxicology and environmental safety, 34(1), 43-55 (1996-06-01)
Mature male rainbow trout (Oncorhynchus mykiss) were exposed for 28 days to 0, 1, 5, and 20 micrograms/liter disulfoton, i.e., to concentrations well below any macroscopically visible effect due to the primary acute toxic mechanism of acetylcholine esterase inhibition. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service