Skip to Content
Merck
All Photos(1)

Key Documents

32015

Supelco

Endosulfan

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6Cl6O3S
CAS Number:
Molecular Weight:
406.93
Beilstein:
2062338
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

(α + β ~ 2 + 1)

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

ClC1=C(Cl)[C@]2(Cl)C3COS(=O)OCC3[C@@]1(Cl)C2(Cl)Cl

InChI

1S/C9H6Cl6O3S/c10-5-6(11)8(13)4-2-18-19(16)17-1-3(4)7(5,12)9(8,14)15/h3-4H,1-2H2/t3?,4?,7-,8+,19?

InChI key

RDYMFSUJUZBWLH-QTSFFCDASA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Endosulfan, a broad spectrum insecticide, is mostly used to control insects and pest in crops. It consists of two stereochemical isomers namely α-endosulfan and β-endosulfan.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kgotla K Masibi et al.
Materials (Basel, Switzerland), 14(4) (2021-02-10)
This report narrates the successful application of a fabricated novel sensor for the trace detection of endosulfan (EDS). The sensor was made by modifying a glassy-carbon electrode (GCE) with polyaniline (PANI), chemically synthesized antimony oxide nanoparticles (AONPs), acid-functionalized, single-walled carbon
Anke M Tukker et al.
Neurotoxicology, 67, 215-225 (2018-06-18)
Neurotoxicity testing still relies on ethically debated, expensive and time consuming in vivo experiments, which are unsuitable for high-throughput toxicity screening. There is thus a clear need for a rapid in vitro screening strategy that is preferably based on human-derived
Claire Bernardin-Souibgui et al.
Environmental science and pollution research international, 25(25), 24860-24881 (2018-06-23)
The nature and fate of urban contaminants washed by stormwater events and accumulating in a detention basin (DB) were investigated. Relations between bacterial and chemical contaminants of trapped urban sediments, and field parameters were analyzed. Fecal indicators and some pathogens
M Guest et al.
Pesticide biochemistry and physiology, 158, 32-39 (2019-08-06)
Mutations in the GABA-gated chloride channel are associated with resistance to cyclodiene organochlorine and phenyl pyrazole insecticides. The best characterised of these is A301S, which was initially identified in a Dieldrin resistant strain of Drosophila melanogaster. The orthologous mutation has
Edebi N Vaikosen et al.
European journal of mass spectrometry (Chichester, England), 25(4), 362-371 (2018-12-19)
Most environmental analytical methods for the determination of organochlorine pesticides (OCPs) are multi-residual with other organic compounds co-extracted and co-eluted. This has been observed in GC spectra using classical detectors like electron-capture detector (ECD) even after appropriate clean-up. This limitation

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service