439223
Dichloromethane
suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Synonym(s):
Methylene chloride
About This Item
6.83 psi ( 20 °C)
Recommended Products
grade
HPLC grade
Quality Level
vapor density
2.9 (vs air)
vapor pressure
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)
Assay
≥99.9%
form
liquid
autoignition temp.
1223 °F
contains
40-150 ppm amylene as stabilizer
expl. lim.
22 %
technique(s)
HPLC: suitable
impurities
Free halogens, passes test
≤0.0003 meq/g Titr. acid
≤0.02% water
evapn. residue
<0.0003%
color
APHA: ≤10
clear
refractive index
n20/D 1.424 (lit.)
bp
39.8-40 °C (lit.)
mp
−95 °C (lit.)
density
1.325 g/mL at 25 °C (lit.)
λ
H2O reference
UV absorption
λ: 235 nm Amax: 1.00
λ: 240 nm Amax: 0.20
λ: 250 nm Amax: 0.05
λ: 260 nm Amax: 0.02
λ: 340-400 nm Amax: 0.01
suitability
passes test for HPLC
SMILES string
ClCCl
InChI
1S/CH2Cl2/c2-1-3/h1H2
InChI key
YMWUJEATGCHHMB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- To compose ultrasonic baths for cleaning and protection of aluminum coating over atactic-PMMA (poly(methyl methacrylate)) for dielectric studies.[7]
- Preparation of AMBF3 (ammoniomethyltrifluoroborate)-conjugated biomolecules.[8]
- Sample preparation for GC-MS (Gas Chromatography-Mass Spectrometry) analysis.[9]
- Preparation of ammonium acetate solvent mixture in combination with isopropanol, acetonitrile and water for mass spectral analysis of neutral lipids.[10]
- Quantification of glutathione (GSH) and glutathione disulfide (GSSG) in biological samples by HPLC and spectrophotometry. [11]
- As an extractant for 2,2-dithioethyl-4-(2′-pyridyl)-4-butyro-γ-lactone.[12]
Packaging
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
Preparation Note
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 2
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service