Skip to Content
Merck
All Photos(1)

Documents

42944

Sigma-Aldrich

Diphenyl diselenide

purum, ≥97.0% (GC)

Synonym(s):

Phenyl diselenide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SeSeC6H5
CAS Number:
Molecular Weight:
312.13
Beilstein:
2047179
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥97.0% (GC)

mp

59-61 °C (lit.)
60-63 °C

SMILES string

[Se]([Se]c1ccccc1)c2ccccc2

InChI

1S/C12H10Se2/c1-3-7-11(8-4-1)13-14-12-9-5-2-6-10-12/h1-10H

InChI key

YWWZCHLUQSHMCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diphenyl diselenide (Ph2Se2) is an organoselenium compound. Its crystalline structure, toxicokinetic properties, antioxidant and anti-inflammatory activities have been investigated. Its ability to reverse the oxidative brain damage and mitochondrial dysfunction induced by acetaminophen has been studied in mice. It reacts with thallium (Tl) to form TI (SePh). It participates in a four-component radical coupling to form substituted cyclopentanes.

Application

Diphenyl diselenide (Ph2Se2) may be used in the following studies:
  • methoxyselenenylation of alkenes
  • dihydroxylation of double bonds
  • hydrothiolation of terminal alkynes
It may be used in the synthesis of the following:
  • unsymmetrical diorganyl selenides
  • 1-(phenylselenomethyl)vinyl selenides
  • allylic phenyl selenides

Other Notes

Reagent for introducing the phenylselenyl group, used for elimination and oxidation reactions, review

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly Selective Sequential Addition and Cyclization Reactions Involving Diphenyl Diselenide, an Alkyne, and Alkenes under Visible-Light Irradiation.
Tsuchii K, et al.
Angewandte Chemie (International Edition in English), 42(30), 3490-3493 (2003)
Eco-Friendly Olefin Dihydroxylation Catalyzed by Diphenyl Diselenide.
Santoro S, et al.
Advanced Synthesis & Catalysis, 350(18), 2881-2884 (2008)
The crystal structure of diphenyl diselenide.
Marsh RE.
Acta Crystallographica, 5(4), 458-462 (1952)
Hydrothiolation of terminal alkynes with diaryl disulfides and diphenyl diselenide: selective synthesis of (Z)-1-alkenyl sulfides and selenides.
Wang ZL, et al.
Tetrahedron, 64(47), 10670-10675 (2008)
Reactions of some main group metals with diphenyl disulphide and diphenyl diselenide.
Kumar R, et al.
J. Chem. Soc., Dalton Trans., 4, 1045-1047 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service