Skip to Content
Merck
All Photos(1)

Key Documents

36125

Supelco

Chloroneb

PESTANAL®, analytical standard

Synonym(s):

2,5-Dichlorohydroquinone dimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8Cl2O2
CAS Number:
Molecular Weight:
207.05
Beilstein:
1952749
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1cc(Cl)c(OC)cc1Cl

InChI

1S/C8H8Cl2O2/c1-11-7-3-6(10)8(12-2)4-5(7)9/h3-4H,1-2H3

InChI key

PFIADAMVCJPXSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroneb is a moderate and systemic fungicide specific to Phytophthora, Pythium, Botrytis and Mucor.

Application

Chloroneb may be used as an analytical reference standard for the determination of the analyte in honey, water, fruits and vegetables by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dispersive liquid--liquid microextraction for the determination of organochlorine pesticides residues in honey by gas chromatography-electron capture and ion trap mass spectrometric detection.
Zacharis CK, et al.
Food Chemistry, 134(3), 1665-1672 (2012)
Determination of pesticides in water by capillary gas chromatography with splitless injection of large sample volumes.
Suzuki T, et al.
Journal of Chromatography A, 662(1), 139-146 (1994)
Determination of multiple pesticides in fruits and vegetables using a modified quick, easy, cheap, effective, rugged and safe method with magnetic nanoparticles and gas chromatography tandem mass spectrometry.
Li YF, et al.
Journal of Chromatography A, 1361(1), 77-87 (2014)
M A Ramesh et al.
Fungal genetics and biology : FG & B, 32(3), 183-193 (2001-05-10)
The cAMP signal transduction pathway mediates the switch between yeast-like and filamentous growth and influences both sexual development and pathogenicity in the smut fungus Ustilago maydis. Signaling via cAMP may also play a role in fungicide resistance in U. maydis.
Degradation of chloroneb, triadimefon, and vinclozolin in soil, thatch, and grass clippings.
E K Frederick et al.
Bulletin of environmental contamination and toxicology, 53(4), 536-542 (1994-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service