Skip to Content
Merck
All Photos(1)

Key Documents

36677

Supelco

Dicofol

PESTANAL®

Synonym(s):

2,2,2-Trichloro-1,1-bis(4-chlorophenyl)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9Cl5O
CAS Number:
Molecular Weight:
370.49
Beilstein:
1886299
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

OC(c1ccc(Cl)cc1)(c2ccc(Cl)cc2)C(Cl)(Cl)Cl

InChI

1S/C14H9Cl5O/c15-11-5-1-9(2-6-11)13(20,14(17,18)19)10-3-7-12(16)8-4-10/h1-8,20H

InChI key

UOAMTSKGCBMZTC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dicofol is a non-systemic insecticide used for a variety of crops. It can be synthesized from dichlorodiphenyltrichloroethane(DDT) via chlorination followed by hydrolysis.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

"Contents and sources of DDT impurities in dicofol formulations in Turkey"
Turgut C, et al.
Environmental Science and Pollution Research International, 16(02), 214-217 (2009)
"Estrogenic activity of dicofol with the human estrogen receptor: Isomer-and enantiomer-specific implications"
Hoekstra.FP, et al.
Chemosphere, 64(01), 174- 177 (2006)
Yukiko Fujii et al.
Chemosphere, 82(1), 25-31 (2010-11-06)
Previously, we demonstrated that the concentrations of DDTs were greater in breast milk collected from Chinese mothers than from Japanese and Korean mothers. To investigate dicofol as a possible source of the DDTs in human breast milk, we collected breast
Xinglun Yang et al.
Journal of hazardous materials, 150(1), 92-98 (2007-06-02)
Dicofol with high impurity of DDT compounds is still widely used in agricultural practice such as cotton cultivation and becomes an important source of DDT pollution in China. In this study, investigations on the DDT residues in cotton fields from
S El-Amrani et al.
The Science of the total environment, 425, 184-190 (2012-04-03)
The feasibility of a bioaccumulation test based on the use of zebrafish eleutheroembryos as an alternative to adult-individual-based approaches for REACH application has been evaluated for three test compounds, chlorpyrifos, dicofol and atrazine. Following the OECD 305 guidelines, zebrafish eleutheroembryos

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service