Skip to Content
Merck
All Photos(1)

Key Documents

34218

Supelco

Cefuroxime

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N4O8S
CAS Number:
Molecular Weight:
424.39
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

[H][C@]12SCC(COC(N)=O)=C(N1C(=O)[C@H]2NC(=O)\C(=N/OC)c3ccco3)C(O)=O

InChI

1S/C16H16N4O8S/c1-26-19-9(8-3-2-4-27-8)12(21)18-10-13(22)20-11(15(23)24)7(5-28-16(17)25)6-29-14(10)20/h2-4,10,14H,5-6H2,1H3,(H2,17,25)(H,18,21)(H,23,24)/b19-9-/t10-,14-/m1/s1

InChI key

JFPVXVDWJQMJEE-IZRZKJBUSA-N

General description

Cefuroxime is a new broad-spectrum cephalosporin antibiotic, with increased stability to β-lactamases. It is actively used against gram-positive organisms, including penicillinase-producing staphylococci and non-β-lactamase-producing gram-negative bacteria.
Chemical structure: ß-lactam

Application

Cefuroxime may be used as a reference standard in the determination of cefuroxime in plasma samples using high-performance liquid chromatography (HPLC).(3)
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cefuroxime, a new cephalosporin antibiotic: activity in vitro.
O'Callaghan HC, et al.
Antimicrobial Agents and Chemotherapy, 9(3), 511-519 (1976)
Simon Farnebo et al.
Biomedicines, 8(10) (2020-10-16)
 Uncomplicated healing of grafts for tendon reconstruction remains an unsolved problem in hand surgery. Results are limited by adhesion formation and decreased strength properties, especially within the tight fibro-osseous sheath of the digits. This is especially problematic when an extra
Dandan He et al.
Antimicrobial agents and chemotherapy, 60(10), 6084-6090 (2016-08-03)
Clinical isolates producing hybrid CTX-M β-lactamases, presumably due to recombination between the blaCTX-M-15 and blaCTX-M-14 elements, have emerged in recent years. Among the hybrid enzymes, CTX-M-64 and CTX-M-14 display the most significant difference in catalytic activity. This study aims to
Grigorios Tsigkas et al.
Hypertension research : official journal of the Japanese Society of Hypertension, 39(12), 840-847 (2016-06-24)
The purpose of this paper is to describe a novel minimally invasive endovascular model of progressive myocardial hypertrophy in rabbits as an experimental protocol of hypertrophic cardiomyopathy. Nine New Zealand White rabbits underwent transauricular aortic catheterization. Under fluoroscopy a bare
Cefuroxime: human pharmacokinetics.
Foord DR.
Antimicrobial Agents and Chemotherapy, 9(5), 741-747 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service