Skip to Content
Merck
All Photos(2)

Documents

34012

Supelco

Fluoxetine hydrochloride

VETRANAL®, analytical standard

Synonym(s):

(±)-N-Methyl-γ-[4-(trifluoromethyl)phenoxy]benzenepropanamine hydrochloride, LY-110,140 hydrochloride, Prozac®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18F3NO · HCl
CAS Number:
Molecular Weight:
345.79
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CNCCC(C1=CC=CC=C1)OC2=CC=C(C(F)(F)F)C=C2.[H]Cl

InChI

1S/C17H18F3NO.ClH/c1-21-12-11-16(13-5-3-2-4-6-13)22-15-9-7-14(8-10-15)17(18,19)20;/h2-10,16,21H,11-12H2,1H3;1H

InChI key

GIYXAJPCNFJEHY-UHFFFAOYSA-N

Gene Information

human ... SLC6A4(6532)

Looking for similar products? Visit Product Comparison Guide

General description

Fluoxetine hydrochloride is an antidepressant drug and a selective serotonin reuptake inhibitor, which is widely used for the treatment of major depressive disorders, obsessive-compulsive disorder and panic fits.

Application

Fluoxetine hydrochloride may be used as an analytical reference standard for the quantification of the analyte in biological fluids using various chromatography techniques.
Fluoxetine hydrochloride may be used as an antidepressant, since it can selectively inhibit the serotonin uptake in the brain. It may also be used in the management of a variety of psychiatric and metabolic derangements.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Selective serotonin reuptake inhibitor; antidepressant.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

Sold under license from Eli Lilly and Company.
Prozac is a registered trademark of Eli Lilly and Co.
VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study of the interaction between fluoxetine hydrochloride and bovine serum albumin in the imitated physiological conditions by multi-spectroscopic methods
Katrahalli U, et al.
Journal of Luminescence, 130, 211-216 (2010)
Molecular Structure of Fluoxetine Hydrochloride, a Highly Selective Serotonin-Uptake Inhibitor
Robertson.WD, et al.
Journal of Medicinal Chemistry, 31, 185-189 (1988)
LC-MS determination and bioavailability study of loperamide hydrochloride after oral administration of loperamide capsule in human volunteers.
Yu H-J, et al.
Journal of Pharmaceutical and Biomedical Analysis, 36(2), 421-427 (2004)
A rapid HPLC-DAD method for the analysis of fluoxetine and norfluoxetine in plasma from overdose patients.
Sabbioni C, et al.
Journal of Pharmaceutical and Biomedical Analysis, 36(2), 351-356 (2004)
Sean C Godar et al.
Journal of psychiatric research, 56, 1-9 (2014-06-03)
The termination of serotonin (5-hydroxytryptamine, 5-HT) neurotransmission is regulated by its uptake by the 5-HT transporter (5-HTT), as well as its degradation by monoamine oxidase (MAO)-A. MAO-A deficiency results in a wide set of behavioral alterations, including perseverative behaviors and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service