Skip to Content
Merck
All Photos(2)

Key Documents

C7861

Sigma-Aldrich

Citalopram hydrobromide

≥98% (HPLC), powder, serotonin uptake inhibitor

Synonym(s):

1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21FN2O · HBr
CAS Number:
Molecular Weight:
405.30
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Citalopram hydrobromide, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

chloroform: soluble
methanol: soluble

originator

Forest Labs

storage temp.

2-8°C

SMILES string

Br[H].CN(C)CCCC1(OCc2cc(ccc12)C#N)c3ccc(F)cc3

InChI

1S/C20H21FN2O.BrH/c1-23(2)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-24-20;/h4-9,12H,3,10-11,14H2,1-2H3;1H

InChI key

WIHMBLDNRMIGDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Citalopram is a bicyclic phthalene derivative. It initiates serotonergic activity, which produces antidepressant action. Citalopram hydrobromide reverses memory impairment. It is used to treat obsessive compulsive disorder, panic disorder, sociophobia and premenstrual dysphoric disorder.

Application

Citalopram hydrobromide has been used:
  • to examine its effects on sirt mRNA and mammalian sirtuins (SIRT) expression in mice
  • to monitor body temperature and antidepressant-like behavioral responses in the forced swim test for rats
  • for 5-hydroxytryptamine (5-HT) competition uptake assays

Biochem/physiol Actions

Potent and selective serotonin uptake inhibitor (Ki = 5.4 nM); antidepressant

Features and Benefits

This compound was developed by Forest Labs. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aging and chronic administration of serotonin-selective reuptake inhibitor citalopram upregulate Sirt4 gene expression in the preoptic area of male mice
Dutt Way W, et al.
Frontiers in Genetics, 6(281), 281-281 (2015)
The Guide to Off-label Prescription Drugs: New Uses for FDA-approved Prescription Drugs (2006)
Interactions between whole-body heating and citalopram on body temperature, antidepressant-like behaviour, and neurochemistry in adolescent male rats
Hale MW, et al.
Behavioural Brain Research, 359, 428-439 (2019)
Whole-body hyperthermia and a subthreshold dose of citalopram act synergistically to induce antidepressant-like behavioral responses in adolescent rats
Hale MW, et al.
Progress in Neuro-Psychopharmacology & Biological Psychiatry, 79, 162-168 (2017)
Current Treatments of Obsessive-Compulsive Disorder (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service