Skip to Content
Merck
All Photos(2)

Documents

33969

Supelco

Indoxacarb

PESTANAL®, analytical standard

Synonym(s):

Methyl 7-chloro-2,5-dihydro-2-[N-(methoxycarbonyl)-4-(trifluoromethoxy)anilinocarbonyl]indeno[1,2-e][1,3,4]oxadiazine-4a(3H)carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17ClF3N3O7
CAS Number:
Molecular Weight:
527.83
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

mixture of isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)N(C(=O)N1CO[C@]2(Cc3cc(Cl)ccc3C2=N1)C(=O)OC)c4ccc(OC(F)(F)F)cc4

InChI

1S/C22H17ClF3N3O7/c1-33-18(30)21-10-12-9-13(23)3-8-16(12)17(21)27-28(11-35-21)19(31)29(20(32)34-2)14-4-6-15(7-5-14)36-22(24,25)26/h3-9H,10-11H2,1-2H3/t21-/m0/s1

InChI key

VBCVPMMZEGZULK-NRFANRHFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indoxacarb belongs to oxadiazine class of insecticides, showing broad-spectrum activity against Lepidoptera and other pests. Its mode of action involves blocking of voltage-gated sodium channels and binding to neuronal nicotinic acetylcholine receptors.

Application

Indoxacarb may be used as an analytical reference standard for the quantification of the analyte in foodstuffs of plant and animal origin using gas chromatography with electron capture detection (GC-ECD) and liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B - STOT RE 1

Target Organs

Blood,Nervous system,Heart, Heart,Nervous system,Blood system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khalid Haddi et al.
Scientific reports, 8(1), 16361-16361 (2018-11-07)
Target-site mutations and changes in insect metabolism or behavior are common mechanisms in insecticide-resistant insects. The co-occurrence of such mechanisms in a pest strain is a prominent threat to their management, particularly when alternative compounds are scarce. Pyrethroid resistance among
Orlando Campolo et al.
Scientific reports, 7(1), 13036-13036 (2017-10-14)
The repeated use of conventional synthetic pesticides in crop protection leads to resistance development by pests along with a negative impact on the environment, particularly non-target arthropods. Plant-derived active compounds, such as essential oils (EOs), play a key role in
Franklin Y Quarcoo et al.
Insects, 10(2) (2019-02-02)
Behavioral symptoms and mortality associated with intoxication with insecticides fipronil and indoxacarb were determined in field-collected eastern subterranean termites, Reticulitermes flavipes (Kollar), and Formosan subterranean termites, Coptotermes formosanus Shiraki. Behaviors and mortality were evaluated at three temperatures (16, 22, and
Dariane Souza et al.
Scientific reports, 9(1), 6713-6713 (2019-05-02)
The western corn rootworm (Diabrotica virgifera virgifera LeConte) (WCR) is a major insect pest of corn (Zea mays L.) in the United States (US) and is highly adaptable to multiple management tactics. A low level of WCR field-evolved resistance to
Determination of indoxacarb residue in foodstuffs of plant and animal origin by GC-ECD and LC-MS/MS.
Dun-Ming Xu, et al.
Agricultural sciences in China, 7(10), 128-1234 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service