Skip to Content
Merck
All Photos(2)

Key Documents

33856

Supelco

α-HCH

PESTANAL®, analytical standard

Synonym(s):

α-1,2,3,4,5,6-Hexachlorocyclohexane, α-BHC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6Cl6
CAS Number:
Molecular Weight:
290.83
Beilstein:
7073451
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

156-161 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Cl[C@@H]1[C@H](Cl)[C@@H](Cl)[C@H](Cl)[C@@H](Cl)[C@H]1Cl

InChI

1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4-,5+,6+/m1/s1

InChI key

JLYXXMFPNIAWKQ-SHFUYGGZSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Lourdes Arellano et al.
Environmental science & technology, 45(21), 9268-9275 (2011-09-17)
Persistent organic pollutants (POPs), including polychlorobiphenyls (PCB), endosulfans, hexachlorocyclohexanes (HCHs), hexachlorobenzene (HCB), polybromodiphenyl ethers (PBDEs), and polycyclic aromatic hydrocarbons (PAHs), were analyzed in snowpack samples collected along an altitudinal gradient (1683-2634 meters above sea level) in the High Tatra Mountains
M Pućko et al.
Environmental science & technology, 46(21), 11862-11869 (2012-10-09)
During the summer of 2009, we sampled 14 partially refrozen melt ponds and the top 1 m of old ice in the pond vicinity for α-hexachlorocyclohexane (α-HCH) concentrations and enantiomer fractions (EFs) in the Beaufort Sea. α-HCH concentrations were 3
Chongguo Tian et al.
Environmental science & technology, 46(4), 2220-2227 (2012-01-21)
Atmospheric outflow of α-HCH from China from 1952 to 2009 was investigated using Chinese Gridded Pesticide Emission and Residue Model (ChnGPERM). The model results show that the outflows via the northeast boundary (NEB, longitude 115-135 °E along 55 °N and
Pernilla Carlsson et al.
Journal of environmental monitoring : JEM, 14(5), 1437-1443 (2012-04-07)
A pilot study is presented evaluating selected chlorinated pesticides as chemical tracers for water masses in a sub-Arctic fjord system (Godthåbsfjord, western Greenland). Polyoxymethylene (POM) based passive water samplers were deployed during summer-autumn 2010. The levels of the analysed chlorinated
Sung-Deuk Choi et al.
Environmental science & technology, 45(20), 8834-8841 (2011-09-13)
An understanding of the factors that control the time trends of persistent organic pollutants (POPs) in the environment is required to evaluate the effectiveness of emission reductions and to predict future exposure. Using a regional contaminant fate model, CoZMo-POP 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service