Skip to Content
Merck
All Photos(1)

Key Documents

33377

Supelco

δ-HCH

PESTANAL®, analytical standard

Synonym(s):

δ-1,2,3,4,5,6-Hexachlorocyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6Cl6
CAS Number:
Molecular Weight:
290.83
Beilstein:
1907334
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

mp

136-140 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

Cl[C@@H]1[C@@H](Cl)[C@H](Cl)[C@@H](Cl)[C@H](Cl)[C@@H]1Cl

InChI

1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4+,5-,6-

InChI key

JLYXXMFPNIAWKQ-GPIVLXJGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Jun Wu et al.
Environmental microbiology, 9(9), 2331-2340 (2007-08-10)
Commercial formulations of hexachlorocyclohexane (HCH) consist of a mixture of four isomers, alpha, beta, gamma and delta. All these four isomers are toxic and recalcitrant pollutants. Sphingobium (formerly Sphingomonas) sp. strain BHC-A is able to degrade all four HCH isomers.
E D Buck et al.
The Journal of pharmacology and experimental therapeutics, 289(1), 477-485 (1999-03-23)
Several isomers of hexachlorocyclohexanes (HCHs) have been shown to be toxic to mammals. Previous studies have revealed that the delta isomer (delta-HCH) was particularly potent toward disrupting Ca2+ homeostasis in a variety of excitable and nonexcitable cells and altering contractility
Poonam Sharma et al.
Applied and environmental microbiology, 72(9), 5720-5727 (2006-09-08)
Incubation of resting cells of Sphingobium indicum B90A, Sphingobium japonicum UT26, and Sphingobium francense Sp+ showed that they were able to transform beta- and delta-hexachlorocyclohexane (beta- and delta-HCH, respectively), the most recalcitrant hexachlorocyclohexane isomers, to pentachlorocyclohexanols, but only resting cells
R Calvelo Pereira et al.
Environmental pollution (Barking, Essex : 1987), 155(2), 350-358 (2007-12-26)
This study focuses on the main routes of distribution and accumulation of different hexachlorocyclohexane (HCH) isomers (mainly alpha-, beta-, gamma- and delta-HCH) in a soil-plant-air system. A field assay was carried out with two plant species, Cynara scolymus L. and
Yong Qian et al.
Environmental monitoring and assessment, 116(1-3), 157-167 (2006-06-17)
The contamination of organochlorine pesticides hexachlorocyclohexane (HCH) and Dichlorodiphenyltrichloroethane (DDT) and their eco-environmental assessment in surface sediments from Lake Dongting, the second-largest freshwater lake in China, were studied. Concentrations of summation HCH (=alpha-HCH + beta-HCH + gamma-HCH +delta-HCH) were 0.21-9.59

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service