Skip to Content
Merck
All Photos(1)

Documents

33375

Supelco

Methoprene

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H34O3
CAS Number:
Molecular Weight:
310.47
Beilstein:
1913191
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COC(C)(C)CCCC(C)C\C=C\C(C)=C\C(=O)OC(C)C

InChI

1S/C19H34O3/c1-15(2)22-18(20)14-17(4)11-8-10-16(3)12-9-13-19(5,6)21-7/h8,11,14-16H,9-10,12-13H2,1-7H3/b11-8+,17-14+

InChI key

NFGXHKASABOEEW-LDRANXPESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methoprene is a juvenile hormone analog and an insect regulator, which can be used to control insects by hindering metamorphosis. It can also be used to control public health pests.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Michael W Dryden et al.
Veterinary parasitology, 191(3-4), 340-346 (2012-10-02)
Monitoring of the performance of flea control products under conditions of natural challenge is valuable in assessing continued effectiveness and determining the ongoing relevance of laboratory studies. A multi-clinic, investigator-blinded study was undertaken in client-owned dogs to investigate and compare
Marek Jindra et al.
Annual review of entomology, 58, 181-204 (2012-09-22)
The molecular action of juvenile hormone (JH), a regulator of vital importance to insects, was until recently regarded as a mystery. The past few years have seen an explosion of studies of JH signaling, sparked by a finding that a
Mehlhorn H
Encyclopedia of Parasitology: A-M Volume 1 (2008)
Yuya Kaieda et al.
Developmental biology, 430(1), 166-176 (2017-08-08)
Steroid hormones regulate life stage transitions, allowing animals to appropriately follow a developmental timeline. During insect development, the steroid hormone ecdysone is synthesized and released in a regulated manner by the prothoracic gland (PG) and then hydroxylated to the active
Rajendhran Rajakumar et al.
Science (New York, N.Y.), 335(6064), 79-82 (2012-01-10)
Complex worker caste systems have contributed to the evolutionary success of advanced ant societies; however, little is known about the developmental processes underlying their origin and evolution. We combined hormonal manipulation, gene expression, and phylogenetic analyses with field observations to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service