Skip to Content
Merck
All Photos(1)

Key Documents

33130

Sigma-Aldrich

4-(Dimethylamino)benzaldehyde

puriss. p.a., reag. Ph. Eur., ≥99% (perchloric acid titration)

Synonym(s):

Ehrlich’s reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4CHO
CAS Number:
Molecular Weight:
149.19
Beilstein:
606802
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
reag. Ph. Eur.

Quality Level

grade

puriss. p.a.

Assay

≥99% (perchloric acid titration)

ign. residue

≤0.1% (as SO4)

loss

≤0.5% loss on drying

mp

72-75 °C (lit.)
73-75 °C

solubility

ethanol: 0.1 g/10 mL

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

CN(C)c1ccc(C=O)cc1

InChI

1S/C9H11NO/c1-10(2)9-5-3-8(7-11)4-6-9/h3-7H,1-2H3

InChI key

BGNGWHSBYQYVRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-(Dimethylamino)benzaldehyde is also commonly referred as Ehrlich′s reagent. Its crystal structure has been investigated by X-ray analysis. Its crystals belong to the monoclinic crystal system and exhibits space group P21/n. 4-(Dimethylamino)benzaldehyde exhibits dual fluorescence. Its dipole moment in various excited states has been evaluated using their electronic spectra and Bilot-Kawski theory.

Application

4-(Dimethylamino)benzaldehyde in micelle (sodium dodecyl sulphate) containing medium may be used for the quantification of hydroxyproline in meat samples by spectrophotometric methods.
Forms colored condensation products (Schiff bases) with pyrroles and primary amines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

327.2 °F - closed cup

Flash Point(C)

164 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kawski A, et al.
Chemical Physics Letters, 448(4), 208-212 (2007)
4-(Dimethylamino) benzaldehyde.
Gao B and Zhu J-L.
Acta Crystallographica Section E, Structure Reports Online, 64(7), 1182-1182 (2008)
Determination of pyrrole derivatives and hydroxyproline with 4-(dimethylamino) benzaldehyde in a micellar medium.
Esteve-Romero JS, et al.
Analytical Letters, 27(8), 1557-1567 (1994)
Jun-Min Guo et al.
Journal of agricultural and food chemistry, 58(11), 6556-6561 (2010-05-15)
A simple colorimetric method for the differentiation of indoleacetic acid (IAA) and indolebutyric acid (IBA) in plant samples is described. The color change is based upon the reaction between the auxins and p-(dimethylamino)benzaldehyde (PDAB, Ehrlich reagent) following the electrophilic substitution
F J Hidalgo et al.
Analytical biochemistry, 262(2), 129-136 (1998-09-29)
The Ehrlich reaction was optimized to determine pyrrolized proteins produced as a consequence of lipid peroxidation and oxidative stress. The procedure consisted of the treatment of the modified protein with p-(dimethylamino)benzaldehyde at a controlled acidity and temperature, and the determination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service