Skip to Content
Merck

D2004

4-(Dimethylamino)benzaldehyde

suitable for histochemical demonstration of nitro blue tetrazolium reduction in neutrophils

Synonym(s):

p-Dimethylaminobenzaldehyde, DMAB, Ehrlich’s reagent

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)2NC6H4CHO
CAS Number:
Molecular Weight:
149.19
UNSPSC Code:
12171500
NACRES:
NA.47
PubChem Substance ID:
EC Number:
202-819-0
Beilstein/REAXYS Number:
606802
MDL number:
Assay:
≥99% (TLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99% (TLC)

form

powder

mp

72-75 °C (lit.)

solubility

ethanol: 50 mg/mL

suitability

suitable for histochemical demonstration of nitro blue tetrazolium reduction in neutrophils

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CN(C)c1ccc(C=O)cc1

InChI

1S/C9H11NO/c1-10(2)9-5-3-8(7-11)4-6-9/h3-7H,1-2H3

InChI key

BGNGWHSBYQYVRX-UHFFFAOYSA-N

Application

4-(Dimethylamino)benzaldehyde (DMAB or Ehrlich’s reagent) has been used as a color reagent for the determination of hydroxyproline level.It forms colored condensation products (Schiff bases) with pyrroles and primary amines. DMAB is also suitable as a reagent to develop latent fingermarks on paper surfaces yielding impressions that are both colored and photoluminescent.
Forms colored condensation products (Schiff bases) with pyrroles and primary amines.


Still not finding the right product?

Explore all of our products under 4-(Dimethylamino)benzaldehyde


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1B

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

327.2 °F - closed cup

flash_point_c

164 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library