Skip to Content
Merck
All Photos(1)

Key Documents

32966

Supelco

Metaflumizone

PESTANAL®, analytical standard

Synonym(s):

2′-[2-(4-Cyanophenyl)-1-(α,α,α-trifluoro-m-tolyl)ethylidene]-4-(trifluoromethoxy)carbanilohydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16F6N4O2
CAS Number:
Molecular Weight:
506.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

FC(F)(F)Oc1ccc(NC(=O)N\N=C(/Cc2ccc(cc2)C#N)c3cccc(c3)C(F)(F)F)cc1

InChI

1S/C24H16F6N4O2/c25-23(26,27)18-3-1-2-17(13-18)21(12-15-4-6-16(14-31)7-5-15)33-34-22(35)32-19-8-10-20(11-9-19)36-24(28,29)30/h1-11,13H,12H2,(H2,32,34,35)/b33-21+

InChI key

MIFOMMKAVSCNKQ-QNKGDIEWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

may contain isomers

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Lact. - Repr. 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophie Tyler et al.
Journal of feline medicine and surgery, 21(4), 282-285 (2018-05-24)
The aim of this study was to obtain information regarding the use of flea treatments and owner-reported flea infestation and skin disease. Owners of 1150 cats were recruited into a long-term longitudinal study ('Bristol Cats' study) and asked to complete
Jun Shen et al.
Pesticide biochemistry and physiology, 139, 53-59 (2017-06-10)
The diamondback moth, Plutella xylostella (L.) has developed resistance to many types of insecticides in the field. To study inheritance and fitness cost of metaflumizone resistance, a susceptible strain of diamondback moth was continuously selected with metaflumizone during 37 generations
Jun Shen et al.
Insects, 11(5) (2020-05-21)
The diamondback moth, Plutella xylostella (L.) is an important pest of cruciferous crops worldwide. It has developed resistance to many conventional and novel insecticide classes. Metaflumizone belongs to the new chemical class of semicarbazone insecticides. To delay the development of
Bo Hu et al.
Insect science, 26(2), 199-216 (2017-09-08)
Cytochrome P450 and UDP-glucosyltransferase (UGT) as phase I and phase II metabolism enzymes, respectively, play vital roles in the breakdown of endobiotics and xenobiotics. Insects can increase the expression of detoxification enzymes to cope with the stress from xenobiotics including
Xing-Liang Wang et al.
Insect science, 23(1), 50-58 (2015-04-09)
Indoxacarb and metaflumizone belong to a relatively new class of sodium channel blocker insecticides (SCBIs). Due to intensive use of indoxacarb, field-evolved indoxacarb resistance has been reported in several lepidopteran pests, including the diamondback moth Plutella xylostella, a serious pest

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service