Skip to Content
Merck
All Photos(2)

Key Documents

32068

Supelco

Cyflumetofen

PESTANAL®, analytical standard

Synonym(s):

2-Methoxyethyl (RS)-2-(4-tert-butylphenyl)-2-cyano-3-oxo-3-(α,α,α-trifluoro-o-tolyl)propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H24F3NO4
CAS Number:
Molecular Weight:
447.45
Beilstein:
11332768
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COCCOC(=O)C(C#N)(C(=O)c1ccccc1C(F)(F)F)c2ccc(cc2)C(C)(C)C

InChI

1S/C24H24F3NO4/c1-22(2,3)16-9-11-17(12-10-16)23(15-28,21(30)32-14-13-31-4)20(29)18-7-5-6-8-19(18)24(25,26)27/h5-12H,13-14H2,1-4H3

InChI key

AWSZRJQNBMEZOI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyflumetofen is an acaricide, which facilitates the rapid knockdown of spider mites and phytophagous mites. Its mode of action involves the inhibition of mitochondrial electron transport and is suitable for use in integrated pest management (IPM) systems.

Application

Cyflumetofen may be used as a reference standard for the determination of the analyte in samples of plant and animal origin using ultrahigh-performance liquid chromatography coupled with tandem mass spectrometry (UHPLC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristen Tollerup et al.
Insects, 11(11) (2020-11-14)
Field experiments were conducted in two commercial almond orchards located in the southern San Joaquin Valley during 2016 and 2017 to evaluate a "preventative" strategy to manage spider mites. Pacific mite, Tetranychus pacificus McGregor, was identified as the only mite
Deng Pan et al.
Journal of economic entomology, 113(2), 918-923 (2019-12-11)
The citrus red mite, Panonychus citri (McGregor), is an important spider mite pest in citrus producing areas. Owing to long-term acaricide exposure, resistance has evolved rapidly in recent years. To evaluate the extent of resistance, seven field mite populations sampled
Nena Pavlidi et al.
Insect biochemistry and molecular biology, 80, 101-115 (2016-12-10)
Cyflumetofen is a recently introduced acaricide with a novel mode of action, acting as an inhibitor of complex II of mitochondrial electron transport chain. It is activated by hydrolysis and the resulting de-esterified metabolite is a much stronger inhibitor. Cyflumetofen
Paul E Bergeron et al.
Pest management science, 76(6), 2170-2179 (2020-01-20)
Biological control in conventional agroecosystems involves the integration of chemical and conservation tactics, requiring knowledge of pesticide non-target effects on key natural enemies. Even for natural enemy groups such as predatory mites (Acari: Phytoseiidae), where pesticide non-target effects have been
Simultaneous determination of cyflumetofen and its main metabolite residues in samples of plant and animal origin using multi-walled carbon nanotubes in dispersive solid-phase extraction and ultrahigh performance liquid chromatography-tandem mass spectrometry.
Li M, et al.
Journal of Chromatography A, 1300, 95-103 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service