Skip to Content
Merck
All Photos(2)

Key Documents

32808

Supelco

Metalaxyl-M

PESTANAL®, analytical standard

Synonym(s):

N-(2,6-Dimethylphenyl)-N-(methoxyacetyl)-D-alanine methyl ester, Mefenoxam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO4
CAS Number:
Molecular Weight:
279.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COCC(=O)N([C@H](C)C(=O)OC)c1c(C)cccc1C

InChI

1S/C15H21NO4/c1-10-7-6-8-11(2)14(10)16(13(17)9-19-4)12(3)15(18)20-5/h6-8,12H,9H2,1-5H3/t12-/m1/s1

InChI key

ZQEIXNIJLIKNTD-GFCCVEGCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Metalaxyl-M belongs to the class of phenylamide fungicides.

Application

Metalaxyl-M may be used as an analytical reference standard for the determination of the analyte in:
  • Maize samples and greenhouse tomato samples by acetonitrile-based extraction and dispersive liquid–liquid microextraction (DLLME) followed by gas chromatography–mass spectrometry (GC-MS).
  • Onion samples by matrix solid-phase dispersion (MSPD) followed by liquid chromatography–tandem mass spectrometric detection (LC-MS/MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Monitoring pesticide residues in greenhouse tomato by combining acetonitrile-based extraction with dispersive liquid-liquid microextraction followed by gas-chromatography-mass spectrometry.
Melo A, et al.
Food Chemistry, 135(3), 1071-1077 (2012)
A simple; efficient and environmentally friendly method for the extraction of pesticides from onion by matrix solid-phase dispersion with liquid chromatography-tandem mass spectrometric detection.
Rodrigues SA, et al.
Analytica Chimica Acta, 678(1), 82-89 (2010)
M P B Mourão et al.
Journal of chromatography. A, 1485, 1-7 (2017-01-29)
In this work, polyurethane-based (PU) devices having cylindrical geometry soaked with suitable organic solvents are proposed for microextraction analysis. This novel analytical approach (PU microextraction; PUμE) operates under the floating sampling technology for extraction, followed by mechanical compression using a
Ananya Prova et al.
The plant pathology journal, 34(5), 367-380 (2018-10-30)
Stems and pods of hyacinth bean cultivated in a farmer's field in Gazipur District, Bangladesh, were found rotted in nearly 5% hyacinth bean plants. A fungus having fluffy mycelium and large sclerotia was isolated from affected tissues. Combined results of
Zachary A Noel et al.
Phytopathology, 109(9), 1628-1637 (2019-04-25)
Current methods to quantitatively assess fungicide sensitivity for a diverse range of oomycetes are slow and labor intensive. Microtiter-based assays can be used to increase throughput. However, many factors can affect their quality and reproducibility. Therefore, efficient and reliable methods

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service