Skip to Content
Merck
All Photos(2)

Key Documents

32064

Supelco

Penthiopyrad

PESTANAL®, analytical standard

Synonym(s):

(RS)-N-[2-(1,3-Dimethylbutyl)-3-thienyl]-1-methyl-3-(trifluoromethyl)pyrazole-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20F3N3OS
CAS Number:
Molecular Weight:
359.41
Beilstein:
11343466
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

CC(C)CC(C)c1sccc1NC(=O)c2cn(C)nc2C(F)(F)F

InChI

1S/C16H20F3N3OS/c1-9(2)7-10(3)13-12(5-6-24-13)20-15(23)11-8-22(4)21-14(11)16(17,18)19/h5-6,8-10H,7H2,1-4H3,(H,20,23)

InChI key

PFFIDZXUXFLSSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Penthiopyrad is a carboxanilide fungicide, mostly used against basidiomycetes and ascomycetes.1

Application

Penthiopyrad may have been used as analytical standard in UPLC–MS–MS analysis for determination of succinate-dehydrogenase-inhibitor (SDHI) fungicide residues from fruits and vegetable samples.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinhua Gan et al.
Food chemistry, 207, 195-204 (2016-04-16)
A multiresidue method for the determination of organofluorine pesticides in fish tissue samples was developed and optimized. This method is based on a cleanup step of the extracted samples with acetonitrile, and carried out by solid phase extraction (SPE) with
Determination of succinate-dehydrogenase-inhibitor fungicide residues in fruits and vegetables by liquid chromatography?tandem mass spectrometry.
Abad-Fuentes, Antonio, et al.
Analytical and Bioanalytical Chemistry, 407.14, 4207-4211 (2015)
Antonios Zambounis et al.
Archives of microbiology, 202(9), 2391-2400 (2020-06-27)
An amplicon metagenomic approach based on the ITS1 region of fungal rDNA was employed to identify the composition of fungal communities associated with diseases of pear fruits during postharvest storage. The sampled fruits were harvested at an orchard using routine
Hyun H Noh et al.
PloS one, 14(4), e0212209-e0212209 (2019-04-10)
The objective of this study was to determine the residual characteristics and to calculate the persistence of the fungicides fluxapyroxad (15.3% suspension concentrate) and penthiopyrad (20% emulsifiable concentrate) on the leaves of greenhouse-cultivated perilla (Perilla frutescens var. japonica Hara). Fluxapyroxad
T R Faske et al.
Journal of nematology, 47(4), 316-321 (2016-03-05)
Fluopyram is a succinate dehydrogenase inhibitor (SDHI) fungicide that is being evaluated as a seed treatment and in-furrow spray at planting on row crops for management of fungal diseases and its effect on plant-parasitic nematodes. Currently, there are no data

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service