Skip to Content
Merck
All Photos(4)

Documents

247561

Sigma-Aldrich

Ammonium citrate dibasic

ACS reagent, 98%

Synonym(s):

Ammonium hydrogencitrate, Citric acid ammonium salt, Diammonium hydrogen citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(CO2H)(CH2CO2NH4)2
CAS Number:
Molecular Weight:
226.18
Beilstein:
4925760
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

1.8 (vs air)

Assay

98%
98.0-103.0% (ACS specification)

form

solid

impurities

oxalate, passes test (lim ~0.05%)
≤0.005% insolubles

ign. residue

≤0.01%

pH

5.2 (20 °C, 50 g/L)

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
heavy metals: ≤5 ppm (by ICP)

SMILES string

N.N.OC(=O)CC(O)(CC(O)=O)C(O)=O

InChI

1S/C6H8O7.2H3N/c7-3(8)1-6(13,5(11)12)2-4(9)10;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);2*1H3

InChI key

YXVFQADLFFNVDS-UHFFFAOYSA-N

Application

Ammonium citrate dibasic can be used as both carbon and nitrogen precursor to synthesize nitrogen-doped fluorescent carbon nanoparticles (CNPs) for the development of sensitive and selective sensors for picric acid detection. Solution form of ammonium citrate can also be used in leaching experiments to extract copper nanoparticles from waste printed circuit boards (WPCBs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Environmentally friendly synthesis of copper nanoparticles from waste printed circuit boards
El-Nasr RS, et al.
Separation and Purification Technology, 230, 115860-115860 (2020)
F Renosto et al.
Journal of bacteriology, 164(2), 674-683 (1985-11-01)
ATP sulfurylases from Penicillium chrysogenum (a mesophile) and from Penicillium duponti (a thermophile) had a native molecular weight of about 440,000 and a subunit molecular weight of about 69,000. (The P. duponti subunit appeared to be a little smaller than
Microwave-assisted ultrafast and facile synthesis of fluorescent carbon nanoparticles from a single precursor: preparation, characterization and their application for the highly selective detection of explosive picric acid
S Xiangcheng, et al.
Journal of Material Chemistry A, 4(11), 4161-4171 (2016)
Influence of surface charge on maximizing solids loading in colloidal processing of alumina.
Singh BP, et al.
Material Matters, 56(4), 475-480 (2002)
Dora Melucci et al.
Annali di chimica, 97(3-4), 141-151 (2007-09-08)
An analytical procedure fit for the simultaneous determination of copper (II), chromium(VI), thallium(I), lead(II), tin(II), antimony(III), and zinc(II) by square wave anodic stripping voltammetry (SWASV) in three interdependent environmental matrices involved in foods and food chain as meals, cereal plants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service