Skip to Content
Merck
All Photos(1)

Documents

22070

Supelco

(+)-Carvone

analytical standard

Synonym(s):

(+)-p-Mentha-6,8-diene 2-one, (S)-5-Isopropenyl-2-methyl-2-cyclohexenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Beilstein:
2042970
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (sum of enantiomers, GC)

optical activity

[α]20/D +61±2°, neat

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.499

bp

228-230 °C (lit.)

density

0.960 g/mL at 20 °C (lit.)

application(s)

agriculture
environmental
food and beverages

format

neat

SMILES string

CC(=C)[C@H]1CC=C(C)C(=O)C1

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)10(11)6-9/h4,9H,1,5-6H2,2-3H3/t9-/m0/s1

InChI key

ULDHMXUKGWMISQ-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

General description

(+)-Carvone (S-carvone), is a monoterpene which has the ability to affect regulation of plant growth and development and microbial activity, it can also prevent sprouting in potato, through inhibition of the induction of phenylalanine ammonia-lyase (PAL) activity, thereby blocking suberisation.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: carum mentha zingiber

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1A

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.1 °F - closed cup

Flash Point(C)

95.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Possible regulatory role of phenylalanine ammonia?lyase in the production of anthocyanins in asparagus (Asparagus officinalis L).
Flores, Francisco B., et al.
Journal of the Science of Food and Agriculture, 85.6, 925-930 (2005)
Haishen Yang et al.
Organic letters, 13(14), 3670-3673 (2011-06-17)
A highly efficient route was developed to synthesize (-)-8-epigrosheimin in four steps from aldehyde 2 based on a substrate-controlled method. The key steps of the synthesis included (1) a stereo- and regioselective allylation addition, (2) an intramolecular translactonization, and (3)
Hee Jin Kim et al.
Organic letters, 13(10), 2682-2685 (2011-04-23)
Selective deoxygenation of allylic alcohol can be successfully carried out by the formation of alkoxyalkyl ether (EE or MOM), followed by Pd(dppe)Cl(2)-catalyzed reduction with LiBHEt(3). (+)-S-Lavandulol has been efficiently synthesized by the application of this protocol to the diol derived
V Y Hatano et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 45(3), 238-243 (2012-02-24)
Lippia alba (Mill.) N.E. Brown (Verbenaceae) is widely used in different regions of Central and South America as a tranquilizer. The plant's anxiolytic properties, however, merit investigation. The present study evaluated the effects of repeated daily (14 days) intraperitoneal (ip)
Raul Conde et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 18(14), 1197-1201 (2011-08-02)
There is no universally accepted and effective prophylaxis of migraine headache episodes. Thus we aimed to investigate the effects of Lippia alba (Mill.) N. E. Brown, an herb with many effects on central nervous system, on pain frequency and intensity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service