Merck
  • Selective deoxygenation of allylic alcohol: stereocontrolled synthesis of lavandulol.
Selective deoxygenation of allylic alcohol: stereocontrolled synthesis of lavandulol.
Organic letters (2011-04-23)
Hee Jin Kim, Liang Su, Heejung Jung, Sangho Koo
ABSTRACT
Selective deoxygenation of allylic alcohol can be successfully carried out by the formation of alkoxyalkyl ether (EE or MOM), followed by Pd(dppe)Cl(2)-catalyzed reduction with LiBHEt(3). (+)-S-Lavandulol has been efficiently synthesized by the application of this protocol to the diol derived from the Pb(OAc)(4)-promoted oxidative ring-opening of (-)-R-carvone. This deoxygenation method is general and selective for allylic alcohols.
Product Description
Supelco
(+)-Carvone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
(+)-Carvone, analytical standard
Supelco
(−)-Carvone, analytical standard
Sigma-Aldrich
(S)-(+)-Carvone, 96%
Sigma-Aldrich
D-Carvone, ≥96%, FG
Sigma-Aldrich
(R)-(−)-Carvone, 98%
Sigma-Aldrich
L-Carvone, ≥97%, FCC, FG
Sigma-Aldrich
L-Carvone, natural, 99%, FG