Skip to Content
Merck
All Photos(1)

Documents

19170

Sigma-Aldrich

3-Buten-1-ol

purum, ≥98.0% (GC)

Synonym(s):

Allylcarbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2CH2OH
CAS Number:
Molecular Weight:
72.11
Beilstein:
1633504
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (GC)

refractive index

n20/D 1.421 (lit.)
n20/D 1.422

bp

112-114 °C (lit.)

density

0.838 g/mL at 25 °C (lit.)

SMILES string

OCCC=C

InChI

1S/C4H8O/c1-2-3-4-5/h2,5H,1,3-4H2

InChI key

ZSPTYLOMNJNZNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Buten-1-ol was used as a starting reagent in asymmetric total synthesis of natural seimatopolide B. It was also used in the synthesis of catalytic bimetallic nanoparticles.

replaced by

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Biologically programmed synthesis of bimetallic nanostructures.
Slocik JM and Naik RR.
Advanced Materials, 18(15), 1988-1992 (2006)
Alternating copolymers of functional alkenes with carbon monoxide.
Kacker S, et al.
Macromolecules, 29(18), 5852-5858 (1996)
Chada Raji Reddy et al.
Organic & biomolecular chemistry, 11(20), 3355-3364 (2013-04-09)
The asymmetric total synthesis of natural seimatopolide B along with its enantiomer is described starting from readily available 5-hexen-1-ol and 3-buten-1-ol. The key steps involved are Jacobson hydrolytic kinetic resolution, proline-catalyzed α-hydroxylation, Yamaguchi esterification and ring-closing metathesis. This asymmetric total

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service