Skip to Content
Merck
All Photos(1)

Key Documents

16797

Supelco

3-Methyl-4-(2,6,6-trimethyl-2-cyclohexen-1-yl)-3-buten-2-one

analytical standard

Synonym(s):

α-Cetone, Isomethyl-α-ionone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22O
CAS Number:
Molecular Weight:
206.32
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.495-1.505

density

0.93 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=O)C(\C)=C\C1C(C)=CCCC1(C)C

InChI

1S/C14H22O/c1-10-7-6-8-14(4,5)13(10)9-11(2)12(3)15/h7,9,13H,6,8H2,1-5H3/b11-9+

InChI key

JRJBVWJSTHECJK-PKNBQFBNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methyl-4-(2,6,6-trimethyl-2-cyclohexen-1-yl)-3-buten-2-one is a fragrance compound primarily utilized in personal care products, including cosmetics, shampoos, deodorants, air fresheners, etc. It is a well-known fragrance allergen known to induce allergic contact dermatitis and rashes.

Application

3-Methyl-4-(2,6,6-trimethyl-2-cyclohexen-1-yl)-3-buten-2-one may be used as an analytical reference standard for the determination of the analyte in perfumes, leave-on cosmetics, commercial scented products, and fragrance raw materials/perfume oils by various chromatography techniques.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

230.5 °F - closed cup

Flash Point(C)

110.3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of fragrance allergens and their dermal sensitization quantitative risk assessment (QRA) in 107 spray perfumes
Soo LD, et al.
Journal of Toxicology and Environmental Health. Part A, 81(22), 1173-1185 (2018)
Development of a solid phase dispersion-pressurized liquid extraction method for the analysis of suspected fragrance allergens in leave-on cosmetics
Lamas JP, et al.
Journal of Chromatography A, 1217(52), 8087-8094 (2010)
Quantitative analysis of the 26 allergens for cosmetic labeling in fragrance raw materials and perfume oils
Leijs H, et al.
Journal of Agricultural and Food Chemistry, 53(14), 5487-5491 (2005)
C Villa et al.
Journal of pharmaceutical and biomedical analysis, 44(3), 755-762 (2007-05-04)
The European legislation on cosmetic products has recently required the declaration of 26 compounds (24 volatile chemicals and 2 natural extracts) on the label of final products when exceeding a stipulated cut-off level. In this work a rapid reliable and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service