Skip to Content
Merck
All Photos(1)

Key Documents

157791

Sigma-Aldrich

Phosphorus trichloride

ReagentPlus®, 99%

Synonym(s):

Phosphorus(III) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PCl3
CAS Number:
Molecular Weight:
137.33
Beilstein:
969177
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
form:
liquid

grade

reagent

Quality Level

vapor density

4.75 (vs air)

vapor pressure

100 mmHg ( 20 °C)
125 mmHg ( 25 °C)
385 mmHg ( 55 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

refractive index

n20/D 1.5148 (lit.)

bp

74-78 °C (lit.)

mp

−112 °C (lit.)

density

1.574 g/mL at 20 °C (lit.)

SMILES string

ClP(Cl)Cl

InChI

1S/Cl3P/c1-4(2)3

InChI key

FAIAAWCVCHQXDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phosphorus trichloride is a slightly yellow or colorless fuming liquid, which finds applications in electrodeposition of metal on rubber and in the preparation of pesticides. It can undergo reaction with chlorine gas to form phosphorus pentachloride.
Rotational spectra of phosphorus trichloride has been studied in the microwave region. Its reaction with unsaturated compounds in the presence of diacetyl peroxide has been reported.

Application

Phosphorus trichloride may be used in the preparation of following:
  • alkylphosphonyl dichlorides
  • benzidronate
  • 4-(dibromomethyl)-substituted benzenes bearing a phosphorus functional group in the side chain
  • dinucleoside phosphorochloridite

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clugston, M. and Flemming, R
Advanced Chemistry (2000)
Synthesis and properties of 4-(dibromomethyl) benzenecarbaldehyde.
Gazizov MB, et al.
Tetrahedron Letters, 57(2), 210-212 (2016)
Yousef Beni et al.
Tetrahedron letters, 56(17), 2247-2250 (2015-12-15)
Dinucleoside phosphorochloridite were synthesized from phosphorus trichloride and three nucleoside analogues, 3'-fluoro-2',3'-dideoxythymidine (FLT), 2',3'-dideoxy-5-fluoro-3'-thiacytidine (FTC), and 2',3'-dideoxy-3'-thiacytidine (3TC), in a multistep synthesis. Polymer-bound
The addition of phosphorus trichloride to olefins.
Kharasch MS, et al.
Journal of the American Chemical Society, 67(10), 1864-1865 (1945)
The microwave spectra and molecular structure of phosphorus and arsenic trichloride.
Kisliuk P and Townes CH.
J. Chem. Phys. , 18(8), 1109-1110 (1950)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service