Skip to Content
Merck
All Photos(1)

Key Documents

14080

Sigma-Aldrich

Succinic acid

purum p.a., ≥99.0% (T)

Synonym(s):

Butanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH2COOH
CAS Number:
Molecular Weight:
118.09
Beilstein:
1754069
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

purum p.a.

Quality Level

Assay

≥99.0% (T)

ign. residue

≤0.05% (as SO4)

bp

235 °C (lit.)

mp

184-186 °C (lit.)
185-188 °C

solubility

methanol: soluble 0.4 g/10 mL

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

functional group

carboxylic acid

SMILES string

OC(=O)CCC(O)=O

InChI

1S/C4H6O4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)

InChI key

KDYFGRWQOYBRFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Succinicacid can be used as an organic additive in the synthesis of:
  • 2-Benzyl substituted isoquinoline N-oxides by rhodium-catalyzed reaction with aryl oximes and secondary propargyl alcohols[1].
  • Spiro-pyrimidine derivatives by cyclization reaction of 5-(2-alkohybenzylidene) barbituric acid[2].
  • Allylselenol via reduction of allyl selenocyanate using lithium aluminum hydride[3].
It can also be used as a reactant to synthesize:
  • Polyesters via catalyst-free polyesterification of glycerol and azelaic acid[4].
  • Poly(glycerol-succinic acid) biodendrimers applicable in specific medical and biotechnological uses[5].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhodium-Catalyzed C- H Activation/Annulation Cascade of Aryl Oximes and Propargyl Alcohols to Isoquinoline N-Oxides
Li Y, et al.
advanced synthesis and catalysis, 363, 3305-3310 (2021)
Chih-Zen Chang et al.
BioMed research international, 2015, 490209-490209 (2015-11-06)
Upregulation of protein kinase B (PKB, also known as Akt) is observed within the cerebral arteries of subarachnoid hemorrhage (SAH) animals. This study is of interest to examine Arctigenin, a potent antioxidant, on endothelial nitric oxide synthase (eNOS) and Akt
Synthesis and characterization of polyesters derived from glycerol, azelaic acid, and succinic acid
Baharu M, et al.
Green Chemistry Letters and Reviews, 8, 31-38 (2015)
Harald Møllendal et al.
The journal of physical chemistry. A, 113(22), 6342-6347 (2009-05-08)
The microwave spectrum of 2-propene-1-selenol, H(2)C=CHCH(2)SeH, has been investigated in the 45-80 GHz spectral range at -30 degrees C. The spectra of six isotopologues of one conformer, which has an anticlinal orientation for the C=C-C-Se chain of atoms and a
Hydride transfer reactions of 5-(2-alkohybenzylidene) barbituric acids: Synthesis of 2, 4, 6-trioxoperhydropyrimidine-5-spiro-3?-chromanes
Krasnov K, et al.
Tetrahedron, 73, 542-549 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service