Skip to Content
Merck
All Photos(5)

Key Documents

S7501

Sigma-Aldrich

Succinic acid

greener alternative

ReagentPlus®, BioRenewable, ≥99.0%

Synonym(s):

Butanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH2COOH
CAS Number:
Molecular Weight:
118.09
Beilstein:
1754069
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71
Pricing and availability is not currently available.

Quality Level

product line

ReagentPlus®

Assay

≥99.0%

greener alternative product characteristics

Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

235 °C (lit.)

mp

184-186 °C (lit.)

solubility

H2O: soluble 1 g/10 mL

greener alternative category

SMILES string

OC(=O)CCC(O)=O

InChI

1S/C4H6O4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)

InChI key

KDYFGRWQOYBRFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is biobased and thus aligns with "Use of Renewable Feedstocks". Click here for more information.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James H Marden et al.
Evolution; international journal of organic evolution, 67(4), 1105-1115 (2013-04-05)
Oxygen conductance to the tissues determines aerobic metabolic performance in most eukaryotes but has cost/benefit tradeoffs. Here we examine in lowland populations of a butterfly a genetic polymorphism affecting oxygen conductance via the hypoxia-inducible factor (HIF) pathway, which senses intracellular
José Manuel Otero et al.
PloS one, 8(1), e54144-e54144 (2013-01-26)
Saccharomyces cerevisiae is the most well characterized eukaryote, the preferred microbial cell factory for the largest industrial biotechnology product (bioethanol), and a robust commerically compatible scaffold to be exploitted for diverse chemical production. Succinic acid is a highly sought after
Hilal Taymaz-Nikerel et al.
Metabolic engineering, 16, 115-129 (2013-02-02)
The interactions between the intracellular metabolome, fluxome and growth rate of Escherichia coli after sudden glycolytic/gluconeogenic substrate shifts are studied based on pulses of different substrates to an aerobic glucose-limited steady-state (dilution rate=0.1h(-1)). After each added glycolytic (glucose) and gluconeogenic
G M Tannahill et al.
Nature, 496(7444), 238-242 (2013-03-29)
Macrophages activated by the Gram-negative bacterial product lipopolysaccharide switch their core metabolism from oxidative phosphorylation to glycolysis. Here we show that inhibition of glycolysis with 2-deoxyglucose suppresses lipopolysaccharide-induced interleukin-1β but not tumour-necrosis factor-α in mouse macrophages. A comprehensive metabolic map
Hanno C Erythropel et al.
Chemosphere, 91(3), 358-365 (2013-01-24)
Phthalate diesters such as di (2-ethylhexyl) phthalate (DEHP) are considered ubiquitous contaminants and are poorly biodegraded in the environment. Moreover, both the parent compound and stable metabolites such as mono (2-ethylhexyl) phthalate (MEHP) are linked to several negative impacts on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service