Skip to Content
Merck
All Photos(3)

Key Documents

114669

Sigma-Aldrich

4-(4-Nitrophenylazo)resorcinol

Dye content 90 %

Synonym(s):

Azoviolet, Magneson I

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4N=NC6H3-1,3-(OH)2
CAS Number:
Molecular Weight:
259.22
Beilstein:
674709
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 90%

mp

195-200 °C (dec.) (lit.)

λmax

432 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1ccc(\N=N\c2ccc(cc2)[N+]([O-])=O)c(O)c1

InChI

1S/C12H9N3O4/c16-10-5-6-11(12(17)7-10)14-13-8-1-3-9(4-2-8)15(18)19/h1-7,16-17H/b14-13+

InChI key

NGPGYVQZGRJHFJ-BUHFOSPRSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

4-(4-Nitrophenylazo) resorcinol, also known as azo violet or magneson I, is a red colored powder. It functions as a spot test reagent and forms blue lake on reaction with Mg(OH)2 in alkaline solution. It is mainly used for determining the presence of magnesium in the solution.
4-(4-Nitrophenylazo)resorcinol is a pH indicator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Gopalan
Inorganic Chemistry for Undergraduates (2009)
Cheng K L
CRC Handbook of Organic Analytical Reagents (1992)
Kuang Lu Cheng, Keihei Ueno, Toshiaki Imamura
CRC Handbook of Organic Analytical Reagents (1992)
National Research Council (U.S.). Food Protection Committee
Food Chemicals Codex (1967)
Highly sensitive determination method for total carbonate in water samples by flow injection analysis coupled with gas-diffusion separation.
Oshima M, et al.
Analytical Sciences, 17, 1285-1290 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service