Skip to Content
Merck
All Photos(2)

Key Documents

07671

Supelco

Anthracene

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Anthraxcene, Paranaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10
CAS Number:
Molecular Weight:
178.23
Colour Index Number:
10790
Beilstein:
1905429
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39011608
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

vapor density

6.15 (vs air)

vapor pressure

1 mmHg ( 145 °C)

product line

TraceCERT®

form

powder or crystals

autoignition temp.

1004 °F

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage condition

under inert gas

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

340 °C (lit.)

mp

210-215 °C (lit.)

solubility

alcohols: soluble
benzene: soluble
chloroform: soluble
hydronaphthalenes: soluble
supercritical carbon dioxide: soluble

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

c1ccc2cc3ccccc3cc2c1

InChI

1S/C14H10/c1-2-6-12-10-14-8-4-3-7-13(14)9-11(12)5-1/h1-10H

InChI key

MWPLVEDNUUSJAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application

Anthracene has been shown to be soluble in a variety of binary and ternary mixtures of cyclohexanone, ethyl acetate, and methanol .

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Payam Payamyar et al.
Chimia, 67(4), 283-285 (2013-08-24)
We describe the challenges involved with extending the limited lateral size of two-dimensional polymers (2DPs). An amphiphilic monomer with three-fold symmetry is chosen to form an ideally tessellated monolayer at the air/water interface. Anthracene [4+4] photo-dimerization is chosen as the
Binjie Sun et al.
Langmuir : the ACS journal of surfaces and colloids, 29(6), 1956-1962 (2013-01-18)
Erythropoietin (EPO) is an attractive protein-unfolding/folding model because of its high degree of unfolding and folding reversibility and intermediate size. Due to its function for regulating red blood cell production by stimulating late erythroid precursor cells, EPO presents obvious values
Hyunjung Lee et al.
Inorganic chemistry, 51(20), 10904-10915 (2012-09-26)
The tendency of a Hg(II) ion to strongly quench fluorescence of potential fluorescent sensors is explored. Fluorescence measurements show the expected order of the chelation-enhanced fluorescence (CHEF) effect of Zn(II) > Cd(II) > Hg(II) ~ Cu(II), which is interpreted as
Alina P Sergeeva et al.
Journal of the American Chemical Society, 134(43), 18065-18073 (2012-10-04)
Clusters of boron atoms exhibit intriguing size-dependent structures and chemical bonding that are different from bulk boron and may lead to new boron-based nanostructures. We report a combined photoelectron spectroscopic and ab initio study of the 22- and 23-atom boron
Jiang-Fei Xu et al.
Organic letters, 15(24), 6148-6151 (2013-11-19)
Dynamic covalent bonds supplied by reversible anthracene dimerization were combined with pillar[5]arene/imidazole host-guest interactions to construct double-dynamic polymers. Heating such polymers (in solution or as a gel) led to depolymerization by dissociation of either the host-guest complexes alone or the

Articles

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service