Skip to Content
Merck
All Photos(2)

Key Documents

05426

Supelco

Acenaphthene

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

1,8-Ethylenenaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10
CAS Number:
Molecular Weight:
154.21
Beilstein:
386081
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

vapor density

5.32 (vs air)

vapor pressure

10 mmHg ( 131 °C)

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

279 °C (lit.)

mp

90-94 °C (lit.)

application(s)

environmental

format

neat

SMILES string

C1Cc2cccc3cccc1c23

InChI

1S/C12H10/c1-3-9-4-2-6-11-8-7-10(5-1)12(9)11/h1-6H,7-8H2

InChI key

CWRYPZZKDGJXCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ningjing Hu et al.
Environmental science and pollution research international, 18(2), 163-172 (2010-06-26)
The levels and possible sources of 16 polycyclic aromatic hydrocarbons (PAHs) were investigated in the surface sediments of Liaodong Bay, Bohai Sea, China. The sum of 16 PAHs (∑PAH(16)) concentrations varied from 144.5 to 291.7 ng/g, with a mean value
Yanzheng Gao et al.
Journal of environmental quality, 40(2), 653-656 (2011-04-28)
The subcellular process and distribution of polycyclic aromatic hydrocarbons (PAHs) in arbuscular mycorrhizal plants remains to be elucidated. This work used a greenhouse experiment to show that, accompanied by the apoplastic and symplastic water movement through the root, acenaphthene (ACE)
Aurore Vergnoux et al.
Environmental research, 111(2), 193-198 (2010-02-13)
Surface (0-5 cm) and subsurface (-5 to 15 cm) soils from burned forest areas in South of France were analyzed to determine contents of 14 priority polycyclic aromatic hydrocarbons (PAHs) and their distribution profile. The sampling procedure allowed us to
F Mermillod-Blondin et al.
Chemosphere, 72(2), 213-223 (2008-03-11)
In urban area, the accumulation of polluted stormwater sediments (SWS) in retention ponds may be a source of dissolved pollutants and nutrients for the aquatic ecosystems. Our objective was to quantify the influence of the thickness of SWS layer and
Zhipei Qin et al.
Journal of chromatography. A, 1196-1197, 89-95 (2008-04-22)
The characterization of a poly(dimethylsiloxane) (PDMS) thin film for active extraction of some polycyclic aromatic hydrocarbons was investigated in both a 1-L aqueous solution and a flow-through system. The thin film was attached to an electric bench-drill at a constant

Articles

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service