Skip to Content
Merck
All Photos(2)

Key Documents

06613

Supelco

Atenolol-d7

analytical standard

Synonym(s):

4-[2-Hydroxy-3-[(1-methylethyl-d7)amino]propoxy]benzeneacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14D7H15N2O3
CAS Number:
Molecular Weight:
273.38
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥94.0% (HPLC)
≥97.0% (isotopic purity)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

application(s)

clinical testing

format

neat

mass shift

M+7

storage temp.

2-8°C

SMILES string

NC(CC1=CC=C(OCC(O)CNC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C1)=O

InChI

1S/C14H22N2O3/c1-10(2)16-8-12(17)9-19-13-5-3-11(4-6-13)7-14(15)18/h3-6,10,12,16-17H,7-9H2,1-2H3,(H2,15,18)/i1D3,2D3,10D

InChI key

METKIMKYRPQLGS-SVMCCORHSA-N

General description

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706030, T706025

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dennis Bernieh et al.
Journal of pharmaceutical and biomedical analysis, 142, 232-243 (2017-05-20)
Dried blood spot (DBS) sampling was investigated as a means of obtaining micro-volume blood samples for the quantitative analyses of ten commonly UK prescribed cardiovascular drugs as an indicator of medication adherence. An 8mm disc was punched out from each
Francesco Riva et al.
Analytical methods : advancing methods and applications, 13(4), 526-535 (2021-01-16)
Sewage sludge is a by-product of wastewater treatment processes, and may be employed in agriculture as a fertilizer or in forestry for land reclamation. It is an important source of nutrients but its reuse can arouse concern on account of
Ruixue Ma et al.
Environment international, 138, 105657-105657 (2020-04-03)
The present work aimed to discuss the enantiomeric occurrence of chiral pharmaceuticals including 5 parent compounds (PCs) metoprolol, propranolol, atenolol, venlafaxine and fluoxetine as well as 6 of their transformation products (TPs) in surface water in Beijing. Among which, 9
Solfrid Hegstad et al.
Therapeutic drug monitoring, 42(2), 335-340 (2019-10-15)
Therapeutic drug monitoring of antihypertensive drugs is being increasingly used to optimize treatment and to assess nonadherence. Separator gels are often used in blood collection tubes to facilitate serum or plasma separation from other blood constituents before analyses. Drug adsorption
Sahar Dalahmeh et al.
The Science of the total environment, 612, 192-201 (2017-08-30)
This study investigated the potential of biochar filters as a replacement or complement for sand filters for removal of pharmaceutically active compounds (PhACs) from wastewater in onsite sewage facilities (OSSF). Specifically, the study investigated the effects of biodegradation, adsorption and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service