Skip to Content
Merck
All Photos(2)

Documents

32414

Supelco

Trimethoprim-d9

VETRANAL®, analytical standard

Synonym(s):

2,4-Diamino-5-(3,4,5-tri(methoxy-d3)benzyl)pyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14D9H9N4O3
CAS Number:
Molecular Weight:
299.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

mass shift

M+9

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])Oc1cc(Cc2cnc(N)nc2N)cc(OC([2H])([2H])[2H])c1OC([2H])([2H])[2H]

InChI

1S/C14H18N4O3/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-17-14(16)18-13(9)15/h5-7H,4H2,1-3H3,(H4,15,16,17,18)/i1D3,2D3,3D3

InChI key

IEDVJHCEMCRBQM-GQALSZNTSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Trimethoprim-d9 may be used as an internal standard in the quantitative determination of pharmaceuticals in incubated soil samples using ultra performance liquid chromatography (UPLC) coupled with a triple quadrupole mass spectrometer.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanna Giebułtowicz et al.
Molecules (Basel, Switzerland), 25(6) (2020-03-28)
In this study, a screening of 26 selected antimicrobials using liquid chromatography coupled to a tandem mass spectrometry method in two Polish wastewater treatment plants and their receiving surface waters was provided. The highest average concentrations of metronidazole (7400 ng/L)
Arnoud de Wilt et al.
Journal of hazardous materials, 304, 84-92 (2015-11-08)
Micropollutant removal in an algal treatment system fed with source separated wastewater streams was studied. Batch experiments with the microalgae Chlorella sorokiniana grown on urine, anaerobically treated black water and synthetic urine were performed to assess the removal of six
Reinhard Oertel et al.
Journal of chromatography. A, 1556, 73-80 (2018-05-12)
A rapid and sensitive hydrophilic interaction liquid chromatography (HILIC)-tandem mass spectrometry method was developed and fully validated for the simultaneous determination of the antidiabetic drug metformin and six further pharmaceuticals (atenolol, gabapentin, levofloxacin, ciprofloxacin, propranolol and trimethoprim) in influent and
Akanksha S Kachhawaha et al.
Journal of AOAC International, 100(3), 592-597 (2017-03-17)
A quick, sensitive multiresidue method was developed for the analysis of 19 multiclass pharmaceuticals and personal care products (PPCPs) in surface water and sewage water. The proposed modified QuEChERS method involved the extraction of water samples (10 mL) with acetonitrile
Sorption and degradation of wastewater-associated non-steroidal anti-inflammatory drugs and antibiotics in soils.
Lin K and Gan J
Chemosphere, 83(3), 240-246 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service