Skip to Content
Merck
All Photos(1)

Documents

03967

Sigma-Aldrich

Starch

from potato, tested according to Ph. Eur.

Synonym(s):

Solani amylum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

biological source

potato

Quality Level

Agency

USP/NF
tested according to Ph. Eur.

form

solid

color

white

pH

5.0-8.0

application(s)

pharmaceutical (small molecule)

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Starch was used for incorporating the erogenous cholesterol ester and [3H] Cholesteryl ethers into rat high density lipoprotein (HDL).It was used for preparing Ac medium for studying the fungal diversity in deep-sea hydrothermal ecosystems.

Biochem/physiol Actions

Starch is a carbohydrate made up of glucose units joined by glycosidic bonds. Undigested part of potato starch is referred as Resistant starch (RS). Resistant starch, RS4 reduces the concentrations of total cholesterol and triglycerides in the serum as well as enhances the concentration of HDL-cholesterol in Wistar rats.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Glass et al.
The Journal of biological chemistry, 260(2), 744-750 (1985-01-25)
The uptake of high-density lipoprotein (HDL)-associated apolipoprotein A-I and cholesterol esters was estimated in 16 tissues of the rat using rat HDL doubly labeled with nondegradable tracers; covalently attached 125I-tyramine-cellobiose traced apo-A-I, and [3H]cholesteryl linoleyl ether traced cholesterol esters. Both
Colin Charnock
Microorganisms, 9(1) (2021-01-07)
Plastic pollution has become one of the most critical environmental issues, as rapidly increasing production, compounded by persistence of plastic wastes in the environment, are outpacing efforts to keep ecosystems plastic-free. A switch to plastics more amenable to microbial attack
Qiang Wu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 928, 106-112 (2013-04-23)
The aqueous two-phase system (ATPS) with polyethylene glycol (PEG) and hydroxypropyl starch (HPS) was evaluated for the extraction of human immunoglobulin G (IgG) from human serum albumin (HSA). The partitions of IgG in PEG/sulphate, PEG/phosphate, PEG/Dextran and PEG/HPS ATPSs were
Should insulin resistance degree be taken into account for assessment of glycemic index?
Jose E Galgani et al.
The American journal of clinical nutrition, 97(4), 902-903 (2013-03-22)
Evolutionary genomics: Detecting selection.
Gregory S Barsh et al.
Nature, 495(7441), 325-326 (2013-03-23)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service