Skip to Content
Merck
All Photos(1)

Key Documents

03960590

Epigallocatechin

primary reference standard

Synonym(s):

(−)-Epigallocatechin, (−)-cis-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol, (−)-cis-3,3′,4′,5,5′,7-Hexahydroxyflavane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O7
CAS Number:
Molecular Weight:
306.27
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

food and beverages

storage temp.

−20°C

SMILES string

O[C@@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3cc(O)c(O)c(O)c3

InChI

1S/C15H14O7/c16-7-3-9(17)8-5-12(20)15(22-13(8)4-7)6-1-10(18)14(21)11(19)2-6/h1-4,12,15-21H,5H2/t12-,15-/m1/s1

InChI key

XMOCLSLCDHWDHP-IUODEOHRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Biochem/physiol Actions

Green tea polyphenol. Anticancer agent. Lower antioxidant activity than EGCG.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongyu Wang et al.
Journal of agricultural and food chemistry, 60(12), 3098-3104 (2012-03-08)
Proanthocyanidins in unripe Manilkara zapota (chiku) were isolated using solvent extraction followed by Sephadex LH-20 fractionation with a yield of 0.9%. HPLC analysis using a diol column revealed well-resolved oligomers ranging from dimer to hexamer. The majority of the proanthocyanidins
Yoo-Hyun Lee et al.
International journal of molecular medicine, 30(1), 69-74 (2012-04-17)
Manipulating acetylation status of key gene targets is likely to be crucial for effective cancer therapy. In this study, we utilized green tea catechins, epicatechin (EC), epigallocatechin (EGC) and epigallocatechin-3-gallate (EGCG) to examine the regulation of androgen receptor acetylation in
Yi-Bin Zhou et al.
Journal of agricultural and food chemistry, 60(7), 1619-1627 (2012-01-10)
Metabolic profiles of broiler chickens were examined after the ingestion of green tea, tea polyphenols, and (-)-epigallocatechin-3-gallate (EGCG). Solid-phase extraction of serum and litters yielded free catechins and their metabolites, which were then identified and quantified by liquid chromatography-tandem mass
Enantioselective total syntheses of (+)-gallocatechin, (-)-epigallocatechin, and 8-C-ascorbyl-(-)-epigallocatechin.
Guang Lin et al.
Chemistry, an Asian journal, 8(4), 700-704 (2013-01-31)
K Koga et al.
Journal of food science, 78(1), H105-H111 (2013-01-03)
The pancreatic lipase inhibitory (PLI) activity of leaf extracts (aqueous, 60 and 99.8 (v/v)% EtOH) of Salacia reticulata Wight, referred to "Kothala himbutu" (KT) in Singhalese, was compared with that of KT stem extracts. Evaporated residue contents and PLI activity

Articles

Validated HPLC method (based on Ph. Eur. Monograph 2668) for quantifying catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e column with advantages in terms of backpressure and matrix robustness.

Validated HPLC method (based on Ph. Eur. Monograph 2668) for quantifying catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e column with advantages in terms of backpressure and matrix robustness.

Validated HPLC method (based on Ph. Eur. Monograph 2668) for quantifying catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e column with advantages in terms of backpressure and matrix robustness.

Validated HPLC method (based on Ph. Eur. Monograph 2668) for quantifying catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e column with advantages in terms of backpressure and matrix robustness.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service