Skip to Content
Merck
All Photos(1)

Documents

E1753

Sigma-Aldrich

(−)-Epicatechin

≥90% (HPLC)

Synonym(s):

(−)-cis-3,3′,4′,5,7-Pentahydroxyflavane, (2R,3R)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O6
CAS Number:
Molecular Weight:
290.27
Beilstein:
92760
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

powder

mp

240 °C (dec.) (lit.)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

O[C@@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3ccc(O)c(O)c3

InChI

1S/C15H14O6/c16-8-4-11(18)9-6-13(20)15(21-14(9)5-8)7-1-2-10(17)12(19)3-7/h1-5,13,15-20H,6H2/t13-,15-/m1/s1

InChI key

PFTAWBLQPZVEMU-UKRRQHHQSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

(−)-Epicatechin (EC) belongs to the group of flavanols and is abundantly present in cacao and cacao products. The chemical structure of EC includes an oxygenated heterocycle with a 4-hydroxyl group linked with two aromatic rings.

Application

(−)-Epicatechin (EC) has been used:
  • as an antioxidant to downregulate nicotinamide adenine dinucleotide phosphate hydrogen (NADPH) oxidase and improve insulin sensitivity in in vivo (high fat-fed mice) and in vitro (HepG2 cells) studies
  • to study its effect on blood pressure, nitric oxide synthase (NOS) activity and anxiety-like behavior on borderline hypertensive rat (BHR) models
  • to test its effect on neuronal survival in the hippocampus of middle-aged rats on swimming training, under oxidative stress
  • to study its antioxidant activity in dark chocolate
  • as a characteristic compound in polyphenols analysis

Biochem/physiol Actions

Epicatechin enhances the skeletal muscle structure in heart failure patients and minimizes cardiometabolic risks. It can be used for treating sarcopenia. Epicatechin reduces myostatin and β-galactosidase and increases the level of markers of muscle growth.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Abhijit et al.
Experimental gerontology, 101, 101-112 (2017-11-28)
The present study explored the effects of swimming training and grape seed proanthocyanidin extract (GSPE) on neuronal survival in the hippocampus (HC) of middle-aged rats along with oxidative stress (OS) parameters. Further, the bioavailability of the GSPE, catechin, epicatechin and
Effects of (-)-epicatechin on molecular modulators of skeletal muscle growth and differentiation
Gutierrez-Salmean G, et al.
The Journal of Nutritional Biochemistry, 25(1), 91-94 (2014)
Effect of dark chocolate on plasma epicatechin levels, DNA resistance to oxidative stress and total antioxidant activity in healthy subjects
Spadafranca A, et al.
The British Journal of Nutrition, 103(7), 1008-1014 (2010)
Michal Kluknavsky et al.
Antioxidants (Basel, Switzerland), 9(2) (2020-01-26)
This study investigated the effects of (-)-epicatechin (Epi) in young male borderline hypertensive rats (BHR) during two weeks of treatment (Epi group, 100 mg/kg/day p.o.) and two weeks post treatment (PE group). Epi reduced blood pressure (BP), which persisted for
Joseph Shay et al.
Oxidative medicine and cellular longevity, 2015, 181260-181260 (2015-07-17)
With recent insight into the mechanisms involved in diseases, such as cardiovascular disease, cancer, stroke, neurodegenerative diseases, and diabetes, more efficient modes of treatment are now being assessed. Traditional medicine including the use of natural products is widely practiced around

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service