Skip to Content
Merck
All Photos(2)

Documents

03826

Supelco

Calcium formate

Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Formic acid calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HCOO)2Ca
CAS Number:
Molecular Weight:
130.11
Beilstein:
3624099
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E238
NACRES:
NA.24

grade

Standard for quantitative NMR
certified reference material
TraceCERT®

Quality Level

description

qNMR Standard for D2O (~7.5 - 8.5 ppm)

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage condition

under inert gas

technique(s)

gas chromatography (GC): suitable
qNMR: suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical

SMILES string

O=CO[Ca]OC=O

InChI

1S/2CH2O2.Ca/c2*2-1-3;/h2*1H,(H,2,3);/q;;+2/p-2

InChI key

CBOCVOKPQGJKKJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium formate is a water soluble salt, which can be obtained as a by-product in the manufacture of polyhydric alcohol. It can serve as an efficient source of calcium in dietary supplements.
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out our entire range of quantitative NMR standards (qNMR standards)

Other Notes

Chemical Shift: ~ 7.5 - 8.5 ppm (chemical shifts may slightly vary depending on the experimental conditions)
suitable NMR solvents: D2O

Recommended products

Check out ChemisTwin, our brand new online portal for identity confirmation of NMR spectra. A feature for quantitative NMR is in development. Learn more or reach out to us for a free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of calcium formate as an accelerator on the physicochemical and mechanical properties of pozzolanic cement pastes
Heikal M
Cement and Concrete Research, 34, 1051-1056 (2004)
Relative bioavailability of calcium from calcium formate, calcium citrate, and calcium carbonate
Hanzlik.PR, et al.
Journal of Pharmacology and Experimental Therapeutics, 313, 1217-1222 (2005)
Absorption and elimination of formate following oral administration of calcium formate in female human subjects
Hanzlik.PR, et al.
Drug Metabolism and Disposition, 33, 282-286 (2005)
Nicolaas A Verwey et al.
Amyloid : the international journal of experimental and clinical investigation : the official journal of the International Society of Amyloidosis, 20(3), 179-187 (2013-07-09)
Abstract Amyloid β-peptide (Aβ) is a key molecule in Alzheimer's disease (AD). Reliable immunohistochemical (IHC) methods to detect Aβ and Aβ-associated factors (AAF) in brain specimens are needed to determine their role in AD pathophysiology. Formic acid (FA) pre-treatment, which
Grant T Corbett et al.
The Journal of biological chemistry, 288(12), 8299-8312 (2013-02-14)
Neurotrophins, such as brain-derived neurotrophic factor (BDNF) and neurotrophin-3 (NT-3), are believed to be genuine molecular mediators of neuronal growth and homeostatic synapse activity. However, levels of these neurotrophic factors decrease in different brain regions of patients with Alzheimer disease

Articles

The signal ratio of two different protons can be measured with tremendous precision, which enables the generation of certified reference materials for use as qNMR standards.

Learn about ChemisTwin® Portal, an online analytical spectra interpretation tool. Discover its NMR lab benefits, workflow integration, and user responsibilities. Explore dRMs, algorithm accuracy, data formats, subscription possibilities, and more.

Learn about ChemisTwin® Portal, an online analytical spectra interpretation tool. Discover its NMR lab benefits, workflow integration, and user responsibilities. Explore dRMs, algorithm accuracy, data formats, subscription possibilities, and more.

Learn about ChemisTwin® Portal, an online analytical spectra interpretation tool. Discover its NMR lab benefits, workflow integration, and user responsibilities. Explore dRMs, algorithm accuracy, data formats, subscription possibilities, and more.

See All

Related Content

ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.

ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.

ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.

ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service