Skip to Content
Merck
All Photos(1)

Documents

02486

Supelco

Pyridine

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N
CAS Number:
Molecular Weight:
79.10
Beilstein:
103233
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.72 (vs air)

vapor pressure

10 mmHg ( 13.2 °C)
20 mmHg ( 25 °C)

Assay

≥99.9% (GC)

autoignition temp.

899 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

12.4 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.509 (lit.)
n20/D 1.510

bp

115 °C (lit.)

mp

−42 °C (lit.)

density

0.978 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

C1=CN=CC=C1

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H

InChI key

JUJWROOIHBZHMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyridine is a heterocyclic compound and a toxic chemical, which is widely used as a solvent and intermediate in the production of dyes, rubber products, paints, textile water repellents, agricultural chemicals, chemical laboratories, manufacturing of piperidine, etc. It plays an important role in the sensory properties of cigarette smoke and exists as a volatile component in certain food products.

Application

Pyridine may be used as an analytical reference standard for the quantification of the analyte in cigarette smoke using reversed-phase liquid chromatography/electrospray ionization tandem mass spectrometry (RP-HPLC–ESI-MS/MS).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Determination of pyridine, 2-picoline, 4-picoline and quinoline from mainstream cigarette smoke by solid-phase extraction liquid chromatography/electrospray ionization tandem mass spectrometry
Saha S, et al.
Journal of Chromatography A, 1217(3), 307-311 (2010)
Total syntheses of complanadines A and B.
Le Zhao et al.
Angewandte Chemie (International ed. in English), 52(6), 1722-1725 (2013-01-03)
Bobby Happ et al.
Chemical Society reviews, 41(6), 2222-2255 (2011-11-15)
Pyridine-based ligands, such as 2,2'-bipyridine and 1,10-phenanthroline, have gained much interest in the fields of supramolecular chemistry as well as materials science. The appealing optoelectronic properties of their complexes with heavy d(6) transition metal ions, such as Ru(ii), Os(II), Re(I)
Wen Zhou et al.
Inorganic chemistry, 52(5), 2271-2273 (2013-02-20)
The reaction of 2,2'-bipyridine (bpy) with monomeric chromium(II) precursors was used to prepare the S = 1 complexes Cr(tBu-acac)2(bpy) (1) and (η(5)-Cp)(η(1)-Cp)Cr(bpy) (3), as well as the S = 2 compound Cr[N(SiMe3)2]2(bpy) (4). The crystallographically determined bond lengths indicate that
Nathalie Ségaud et al.
Inorganic chemistry, 52(2), 691-700 (2013-01-11)
We report the synthesis, characterization, and solution chemistry of a series of new Fe(II) complexes based on the tetradentate ligand N-methyl-N,N'-bis(2-pyridyl-methyl)-1,2-diaminoethane or the pentadentate ones N,N',N'-tris(2-pyridyl-methyl)-1,2-diaminoethane and N,N',N'-tris(2-pyridyl-methyl)-1,3-diaminopropane, modified by propynyl or methoxyphenyltriazolyl groups on the amino functions. Six of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service