Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyridine
Synonym(s):
Pyridine
Empirical Formula (Hill Notation):
C
5
H
5
N
CAS Number:
110-86-1
Molecular Weight:
79.10
EC Number:
203-809-9
Products
Product Category
solvents (10)
aroma chemicals (1)
building blocks (1)
Brand
Sigma-Aldrich (12)
Supelco (4)
Boiling Point (°C)
Color
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Package Size
Special Grade
ACS reagent (3)
Puriss (3)
Anhydrous (2)
Puriss P.A. (2)
Biotechnology (1)
Fragrance grade (1)
Halal (1)
Kosher (1)
analytical standard (1)
certified reference material (1)
Show More
Technique
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
360570
Pyridine,
ACS reagent, ≥99.0%
Expand
View Pricing
270407
Pyridine,
suitable for HPLC, ≥99.9%
Expand
View Pricing
P57506
Pyridine,
ReagentPlus
®
, ≥99%
Expand
View Pricing
1.09728
Pyridine,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
5.89579
Pyridine,
anhydrous, 99.8%
Expand
View Pricing
W296600
Pyridine,
≥99%
Expand
View Pricing
270970
Pyridine,
anhydrous, 99.8%
Expand
View Pricing
33553
Pyridine,
puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Expand
View Pricing
320498
Pyridine,
ReagentPlus
®
, ≥99%
Expand
View Pricing
1.07462
Pyridine,
EMPLURA
®
Expand
View Pricing
494410
Pyridine,
biotech. grade, ≥99.9%
Expand
View Pricing
PHR1558
Pyridine,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing
16037
Pyridine,
≥99.5% (GC)
Expand
View Pricing
02486
Pyridine,
analytical standard
Expand
View Pricing
33553-M
Pyridine,
puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Expand
View Pricing
P57506-M
Pyridine,
reagent grade, ≥99%
Expand
View Pricing