Skip to Content
Merck
All Photos(1)

Key Documents

00820580

Verbascoside

primary reference standard

Synonym(s):

Acetoside, Kusaginin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H36O15
CAS Number:
Molecular Weight:
624.59
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](OCCc3ccc(O)c(O)c3)O[C@H](CO)[C@H]2OC(=O)\C=C\c4ccc(O)c(O)c4)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C29H36O15/c1-13-22(36)23(37)24(38)29(41-13)44-27-25(39)28(40-9-8-15-3-6-17(32)19(34)11-15)42-20(12-30)26(27)43-21(35)7-4-14-2-5-16(31)18(33)10-14/h2-7,10-11,13,20,22-34,36-39H,8-9,12H2,1H3/b7-4+/t13-,20+,22-,23+,24+,25+,26+,27+,28+,29-/m0/s1

InChI key

FBSKJMQYURKNSU-ZLSOWSIRSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Strandard in the analysis of herbal medicinal products

Biochem/physiol Actions

Phenylethanoid glycoside. Antioxidant and antiproliferative.

Other Notes

This compound is commonly found in plants of the genus: echinacea plantago

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia Vertuani et al.
Molecules (Basel, Switzerland), 16(8), 7068-7080 (2011-08-20)
We here report the results of our investigations carried out on verbascoside, a phenylpropanoid glycoside known for its antioxidant, anti-inflammatory and photoprotective actions. Verbascoside was obtained from Buddleia davidii meristematic cells, obtained in turn using a sustainable biotechnology platform which
Agata Wilczańska-Barska et al.
Biotechnology letters, 34(9), 1757-1763 (2012-05-29)
Hairy root cultures of Scutellaria lateriflora were established using Agrobacterium rhizogenes A4 and produced acteoside (18.5 mg g(-1) dry wt), baicalin (14.5 mg g(-1) dry wt) and wogonoside (12 mg g(-1) dry wt). Yeast extract (50 μg ml(-1)) increased acteoside
Sándor Gonda et al.
Phytochemistry, 86, 127-136 (2012-11-22)
The aim of this study was to test contribution of plant-associated microorganism (PAMs) to metabolite stability/instability in a medicinal plant matrix. Therefore, PAM strains were isolated and identified based on relevant DNA sequences from Plantago lanceolata leaves. Sterile water extracts
Gianluca Campo et al.
Journal of thrombosis and thrombolysis, 34(3), 318-325 (2012-06-23)
Preliminary in vitro and animal studies have shown that verbascoside, a phenolic compound, may have several favourable biological activities, including an influence on endothelial function and on platelet aggregation. We sought to evaluate the effects of verbascoside, biotechnologically produced from
Paola Di Leo Lira et al.
Chemistry & biodiversity, 10(2), 251-261 (2013-02-19)
The aerial parts of lemon verbena (Aloysia citriodora PALÁU) are worldwide used due to their medicinal and aromatic properties. The essential-oil and acteoside contents have been proposed as the main quality markers for their pharmacological and organoleptic features. The northwestern

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service