Skip to Content
Merck
All Photos(1)

Key Documents

RTT-RO

Roche

Terminal Transferase

from Calf Thymus, recombinant, E. coli

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
NACRES:
NA.21
Pricing and availability is not currently available.

recombinant

expressed in E. coli

Quality Level

form

solution

usage

sufficient for 20 reactions (03333566001)
sufficient for 60 reactions (03333574001)

packaging

pkg of 24,000 U (03333574001 [400 U per reaction])
pkg of 8,000 U (03333566001 [400 U per reaction])

manufacturer/tradename

Roche

application(s)

genomic analysis

storage temp.

−20°C

General description

Terminal Transferase catalyzes the template independent addition of deoxy- and dideoxynucleoside triphosphates to the 3′-OH ends of double and single-stranded DNA fragments, and oligonucleotides. Terminal Transferase incorporates digoxigenin-, biotin-, and fluorochrome-labeled deoxy- and dideoxynucleoside triphosphates as well as radioactively labeled deoxy- and dideoxynucleoside triphosphates. The supplied 5x-concentrated reaction buffer allows the optimal tailing of all types of double-stranded DNA ends: blunt ended, with 3′ overhang, or with 5′ overhang. The highest incorporation rates are obtained with 3′ overhangs.

Application

Use terminal transferase to add nucleotides to the 3′-OH ends of double- or single-stranded DNA fragments, for example:
  • Tailing with dNTPs:Addition of homopolymeric tails to DNA fragments[1][2][3]
  • Labeling of double- and single-stranded DNA and oligonucleotides with either radioactive or chemically modified nucleotides (e.g., DIG-dUTP)
3′-end Labeling with ddNTPs:
Labeling of double- and single-stranded DNA and oligonucleotides with either radioactive or chemically modified dideoxynucleotides (e.g., DIG-ddUTP)

Features and Benefits

Incorporation of labeled or modified nucleotides
In addition to standard nucleotides, terminal transferase wlll add radioactive or modified (e.g., digoxigenin-, biotin-, or fluorochrome-labeled) dNTPs or ddNTPs to DNA.

Packaging

1 kit containing 3 components

Quality

Absence of 5′ and 3′ exonucleases, endonucleases, and nicking activities tested according to the current Quality Control procedures.

Principle

Oligonucleotides are enzymatically labeled at their 3′ end using terminal transferase by incorporation of a single digoxigenin-labeled dideoxyuridine-triphosphate. Another way to label oligonucleotides is the addition of a longer nucleotide tail. For the generation of tailed oligonucleotide probes, deoxynucleotides triphosphates are used in a template independent reaction.

Unit Definition

One unit is the enzyme activity that incorporates 1 nMol dTMP into acid-insoluble products within 30 minutes at +37 °C under assay conditions using d(pT)6 as primer. Unit assay conditions: 200 mM Potassium cacodylate, 1 mM CoCl2, 1 mM dTTP, 0.1 OD d(pT)6, 6.25 pmol 3H dTTP in a 120 μl reaction volume.

Unit Assay: Unit assay conditions: 200 mM Potassium cacodylate, 1 mM CoCl2, 1 mM dTTP, 0.1 OD d(pT)6, 6.25 pmol 3H dTTP in a 120 μl reaction volume.

Volume Activity: 400 U/μl

Sample Materials
  • Double- or single-stranded DNA fragments
  • Double- or single-stranded oligonucleotides

Preparation Note

Working solution: Standard Tailing reaction with radioactive nucleotides
Preparation of CoCl2 working solution
Add in a sterile vial 10 μl double dist. water and 15 μl of the supplied 25 mM CoCl2 solution: Final concentration: 15 mM

Preparation of radioactive labeling mix
dATP and dTTP labeling mix: mix 1 Vol. of a 2.5 mM dATP or dTTP solution with 15 volumes of double-distilled water and 4 volumes of α-32P-dATP or α-32P-dTTP (800 Ci/mmol, approx. 30 TBq/mmol).
dGTP and dCTP labeling mix: mix 1 volume of a 2 mM dGTP or dCTP solution with 15 volumes of double-distilled water and 4 volumes of α-32P-dGTP or α-32P-dCTP (800 Ci/mmol, approx. 30 TBq/mmol)

Other Notes

For general laboratory use. Double-stranded DNA may have either blunt-, 3′-protruding, or 5′-protruding ends. However, 3′-protruding ends lead to the highest incorporation rates.TdT requires an oligonucleotide of at least three bases as a primer, and single-stranded DNA is tailed more efficiently than double-stranded.

Kit Components Only

Product No.
Description

  • Terminal Transferase 400 U/μl

  • TdT Reaction Buffer 5x concentrated

  • CoCl<sub>2</sub> Solution 25 mM

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B Inhalation - Repr. 1B

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giselle A Suero-Abreu et al.
Neoplasia (New York, N.Y.), 16(12), 993-1006 (2014-12-17)
Mouse models have increased our understanding of the pathogenesis of medulloblastoma (MB), the most common malignant pediatric brain tumor that often forms in the cerebellum. A major goal of ongoing research is to better understand the early stages of tumorigenesis
Wolfgang Jäger et al.
PloS one, 8(3), e59536-e59536 (2013-04-05)
Orthotopic bladder cancer xenografts are essential for testing novel therapies and molecular manipulations of cell lines in vivo. Current xenografts rely on tumor cell inoculation by intravesical instillation or direct injection into the bladder wall. Instillation is limited by the
Arnaud Augert et al.
Free radical biology & medicine, 65, 969-977 (2013-09-03)
Little is known about the biological functions of the phospholipase A2 receptor (PLA2R1) except that it has the ability to bind a few secreted phospholipases A2 (sPLA2's). We have previously shown that PLA2R1 regulates senescence in normal human cells. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service