Skip to Content
Merck
All Photos(1)

Key Documents

SCC126M

Sigma-Aldrich

SF7761 Human DIPG H3.3-K27M Cell Line

Human

Synonym(s):

Diffuse intrinsic pontine glioma cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81

Product Name

SF7761 Human DIPG H3.3-K27M Cell Line, SF7761 pediatric diffuse intrinsic pontine glioma (DIPG) cell line harbors the histone H3.3 Lys 27-to-methionine (K27M) mutation and can support research and drug development efforts targeting DIPG.

biological source

human

Quality Level

technique(s)

cell culture | mammalian: suitable

General description

Diffuse intrinsic pontine gliomas (DIPG) are highly aggressive and difficult to treat tumors arising in the ventral pons of the brain stem. Despite therapeutic advances, DIPG is incurable and most patients, primarily children, die within 2 years of diagnosis. DIPG is one of the leading causes of death in children with brain tumors .

A somatic mutation of histone H3.3 resulting in a lysine 27 to methionine substitution (H3.3K27M) occurs in 60% of DIPG . In H3.3K27M DIPG patient samples, levels of H3K27 dimethylation (H3K27me2) and trimethylation (H3K27me3) are reduced globally. Expression of H3.3K27M was also shown to be associated with increased levels of H3K27 acetylation (H3K27ac) and recruitment of bromodomain proteins at sites of active transcription . These epigenetic changes are thought to be important factors driving DIPG oncogenesis
SF7761 is a human glioma cell line derived by surgical biopsy from a young female H3.3K27M DIPG patient (5). The tumor cells were immortalized with hTERT (human telomerase ribonucleoprotein reverse transcriptase) using retroviral transduction. SF7761 cells are tumorigenic in athymic rodents with the tumor cells recapitulating the infiltrative growth of human brainstem gliomas (5, 6).

IMPORTANT NOTE: SF7761 are grown as neurospheres in suspension culture. Neurospheres are to be passaged using gentle mechanical trituration when they reach diameters of 200-500 um or through enzymatic dissociation.

Cell Line Description

Cancer Cells

Application

Research Category
Cancer

Oncology
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Quality

• Each vial contains ≥ 200 viable neurospheres.
• Cells are tested negative for HPV-16, HPV-18, Hepatitis A, C, Herpesvirus type 6, 7, 8 and HIV-1 & 2 viruses by PCR.
• Cells are negative for mycoplasma contamination.
• Each lot of cells is genotyped by STR analysis to verify the unique identity of the cell line.

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kui-Ming Chan et al.
Genes & development, 27(9), 985-990 (2013-04-23)
Recent studies have identified a Lys 27-to-methionine (K27M) mutation at one allele of H3F3A, one of the two genes encoding histone H3 variant H3.3, in 60% of high-grade pediatric glioma cases. The median survival of this group of patients after
Kristin M Schroeder et al.
Pediatric research, 75(1-2), 205-209 (2013-11-07)
Diffuse intrinsic pontine glioma (DIPG) is a high-grade glioma that originates in the pons and is seen exclusively in children. Despite numerous efforts to improve treatment, DIPG remains incurable with 90% of children dying within 2 y of diagnosis, making
Rintaro Hashizume et al.
Journal of neuro-oncology, 110(3), 305-313 (2012-09-18)
Diffuse intrinsic pontine gliomas arise almost exclusively in children, and despite advances in treatment, the majority of patients die within 2 years after initial diagnosis. Because of their infiltrative nature and anatomic location in an eloquent area of the brain
Andrea Piunti et al.
Nature medicine, 23(4), 493-500 (2017-03-07)
Diffuse intrinsic pontine glioma (DIPG) is a highly aggressive pediatric brainstem tumor characterized by rapid and uniform patient demise. A heterozygous point mutation of histone H3 occurs in more than 80% of these tumors and results in a lysine-to-methionine substitution
Rintaro Hashizume et al.
Nature medicine, 20(12), 1394-1396 (2014-11-18)
Pediatric brainstem gliomas often harbor oncogenic K27M mutation of histone H3.3. Here we show that GSKJ4 pharmacologic inhibition of K27 demethylase JMJD3 increases cellular H3K27 methylation in K27M tumor cells and demonstrate potent antitumor activity both in vitro against K27M

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service