Skip to Content
Merck
All Photos(1)

Key Documents

PX1840

Supelco

Isopropyl Alcohol

Isopropyl Alcohol

70% Reagent

Synonym(s):

IPA, 70% alcohol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352104
grade:
reagent
vapor pressure:
42 hPa ( 20 °C)
Pricing and availability is not currently available.

grade

reagent

Quality Level

vapor pressure

42 hPa ( 20 °C)

form

liquid

shelf life

36 mo. (from manufacture date)

composition

Isopropyl Alcohol, 70%

color

APHA: ≤10

pH

7 (20 °C)

transition temp

flash point 18 °C

density

0.85 g/cm3 at 20 °C

shipped in

ambient

storage temp.

no temp limit

Application

Isopropyl Alcohol can be used as:
  • A ‘green′ reaction solvent for the preparation of high molecular weight, high strength cycloaliphatic diisocyanate based silicone-urea copolymers.[1]
  • A granulating agent to determine the endpoint for microcrystalline cellulose (MCC-102) using the powder rheometer and thermal analysis.[2]
  • An extraction solvent for the extraction of oil from microalgae that have high polar lipid content.[3]
  • A solvent in the dehydration and paraffinization of tissues.[4]

Analysis Note

Expiration date36 months from mfg dateColor (APHA)10APHA maxFormClear liquidInfrared SpectrumConforms to standardSpecific Gravity at 20/20°C0.872-0.883

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

64.4 °F

Flash Point(C)

18 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

To prepare and characterize microcrystalline cellulose granules using water and isopropyl alcohol as granulating agents and determine its end-point by thermal and rheological tools
Chaudhari SP and Dave RH
Drug Development and Industrial Pharmacy, 41(5), 744-752 (2015)
I V Viktorov et al.
Bulletin of experimental biology and medicine, 136(1), 105-106 (2003-10-10)
Isopropyl alcohol (isopropanol) can be used as a substitute for ethyl alcohol in tissue dehydration during embedding into paraffin and dehydration of stained sections. The use of isopropyl alcohol during paraffinization allows us to exclude treatment with intermediate solvents of
Isopropyl alcohol: an unusual, powerful,`green?solvent for the preparation of silicone-urea copolymers with high urea contents
Yilgor E, et al.
Polymer, 44(26), 7787-7793 (2003)
Oil extraction from microalga Nannochloropsis sp. with isopropyl alcohol
Yao Linxing, et al.
Journal of the American Oil Chemists' Society, 89(12), 2279-2287 (2012)
Smruti P Chaudhari et al.
Drug development and industrial pharmacy, 41(5), 744-752 (2014-03-25)
Microcrystalline cellulose (MCC-102) is one of the most commonly used excipient in the pharmaceutical industry. For this research purpose, authors have developed a different technique to determine the end point for MCC-102 using water and isopropyl alcohol 70% (IPA) as

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. Ohio
    • Review 1
    • Vote 1
    5 out of 5 stars.

    Good customer service!

    In stock and quick turn around. Friendly people to work with.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service