Skip to Content
Merck
All Photos(1)

Key Documents

OP17

Sigma-Aldrich

Anti-c-Fos (Ab-1) Mouse mAb (2G9C3)

liquid, clone 2G9C3, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2G9C3, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

rat, human, mouse

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG2a

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FOS(2353)

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with AG.8653X mouse myeloma cells. Recognizes the ~55 kDa c-fos and ~62 kDa v-fos proteins.
Recognizes both the ~55 kDa c-Fos and ~62 kDa v-Fos proteins. Does not cross-react with the ~39 kDa Jun protein.
This Anti-c-Fos (Ab-1) Mouse mAb (2G9C3) is validated for use in Immunoblotting, Immunocytochemistry, Paraffin Sections for the detection of c-Fos (Ab-1).

Immunogen

Human
a synthetic peptide (KVEQLSPEEEEKRRIRRERNKMAAA) corresponding to amino acids 128-152 of human c-Fos

Application

Immunoblotting (2.5-5 µg/ml)

Immunocytochemistry (2.5-5 µg/ml)

Paraffin Sections (not recommended)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM sodium phosphate buffer, 0.2% gelatin, pH 7.5.

Analysis Note

Positive Control
HeLa cells

Other Notes

Jensen, D.E., et al. 1991. Oncogene6, 1219.
Reiss, M., et al. 1990. Cancer Res.50, 6641.
RouscherIII, F.J., et al. 1988. Science240, 1010.
Sassone-Corsi, P., et al. 1988. Cell54, 553.
Giardina, S.L., et al. 1987. Anal Biochem. 161, 109.
Verma, I.M., et al. 1987. Cancer Res.49, 29.
Verma, I.M., et al. 1987. Cell51, 513.
Muller, R. 1986. Viochem. Biophys. Acta.823, 207.
Verma, P. 1986. Trends Genet.2, 93.
Greenberg, M., et al. 1984. Nature (London)311, 433.
Recognizes both cellular and viral forms of fos. Does not react with the ~39 kDa jun protein. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erin M Johnson-Venkatesh et al.
Development (Cambridge, England), 142(22), 3879-3891 (2015-09-30)
Neuronal activity, including intrinsic neuronal excitability and synaptic transmission, is an essential regulator of brain development. However, how the intrinsic neuronal excitability of distinct neurons affects their integration into developing circuits remains poorly understood. To investigate this problem, we created
Juan Jurado et al.
BMC molecular biology, 8, 83-83 (2007-09-25)
Alternative splicing is a widespread mechanism of gene expression regulation. Previous analyses based on conventional RT-PCR reported the presence of an unspliced c-fos transcript in several mammalian systems. Compared to the well-defined knowledge on the alternative splicing of fosB, the
Gourav Sharma et al.
Frontiers in molecular neuroscience, 15, 943980-943980 (2022-10-11)
The Ubiquitin Proteasome System (UPS) has been shown to regulate neuronal development and synapse formation. Activity-dependent regulation of E3 ligase, a component of the UPS that targets specific proteins for proteasome-mediated degradation, is emerging as a pivotal player for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service