Skip to Content
Merck
All Photos(1)

Key Documents

200R-1

Sigma-Aldrich

Androgen Receptor (SP107) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SP107, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (200R-14)
vial of 0.5 mL concentrate (200R-15)
bottle of 1.0 mL predilute (200R-17)
vial of 1.0 mL concentrate (200R-16)
bottle of 7.0 mL predilute (200R-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200

isotype

IgG

control

prostate

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

General description

As a member of the steroid hormone superfamily, androgen receptor functions within distinctintracellular compartments to mediate cellular growth and transcriptional activation ofandrogen-regulated signaling pathways. Androgen receptor expression is broadly distributedacross a variety of normal tissue types. The anti-androgen receptor may be used as an aid toidentify carcinomas of the breast and prostate.

Quality


IVD

IVD

IVD

RUO

Linkage

Androgen Receptor Positive Control Slides, Product No. 200S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura C Collins et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 24(7), 924-931 (2011-05-10)
Previous studies have demonstrated that androgen receptor is expressed in many breast cancers, but its expression in relation to the various breast cancer subtypes as defined by molecular profiling has not been studied in detail. We constructed tissue microarrays from
A K Roy et al.
Annals of the New York Academy of Sciences, 949, 44-57 (2002-01-25)
Androgens are C-19 steroids secreted primarily from the testes and adrenals that play a critical role in reproduction. Reproductive functions of androgens are mediated through coordination of diverse physiological processes ranging from brain functions to specific cell proliferation and apoptosis.
Carlos Cordon-Cardo et al.
The Journal of clinical investigation, 117(7), 1876-1883 (2007-06-09)
We have developed an integrated, multidisciplinary methodology, termed systems pathology, to generate highly accurate predictive tools for complex diseases, using prostate cancer for the prototype. To predict the recurrence of prostate cancer following radical prostatectomy, defined by rising serum prostate-specific
R S Carroll et al.
Journal of neurosurgery, 82(3), 453-460 (1995-03-01)
The predominance of meningiomas in females, the accelerated growth of these tumors during the luteal phase of the menstrual cycle and during pregnancy, and the association between meningiomas and breast cancer have led to a number of studies examining the
Claire Bléchet et al.
Virchows Archiv : an international journal of pathology, 450(4), 433-439 (2007-03-03)
Previous studies have shown that C cells are twice as numerous in male than in female thyroids and that C cell hyperplasia (CCH) is much more frequent in men. These findings suggest regulation involving sex steroid hormones through the expression

Articles

IHC antibodies enhance dermatopathology beyond H&E stained slides, improving techniques and applications for dermatological research.

IHC antibodies enhance dermatopathology beyond H&E stained slides, improving techniques and applications for dermatological research.

IHC antibodies enhance dermatopathology beyond H&E stained slides, improving techniques and applications for dermatological research.

IHC antibodies enhance dermatopathology beyond H&E stained slides, improving techniques and applications for dermatological research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service