Skip to Content
Merck
All Photos(1)

Key Documents

MXQP0WUS1

Millipore

Power Supply for Milliflex® Quantum Reader

Milliflex® Quantum Rapid Detection System power supply for United States

Synonym(s):

For rapid detection of microbial contamination in filterable samples, suitable for bioburden testing, Power Supply- United States

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
eCl@ss:
32160606
NACRES:
NB.79

Pricing and availability is not currently available.

material

304 stainless steel plate
polypropylene funnel(s)

manufacturer/tradename

Milliflex®

application(s)

pharmaceutical

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
PZ0184PZ0333SML2809
WNK463 ≥98% (HPLC)

SML2809

WNK463

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

2-8°C

solubility

DMSO: >30 mg/mL

solubility

DMSO: 20 mg/mL

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

color

white to off-white

color

white to beige

color

white to beige

color

white to beige

General description

Power supply is an additional accessory for Milliflex® Quantum Rapid Detection System. Milliflex® Quantum System is a rapid fluorescence-based technology for quantitative detection of contaminants over a broad range of filterable matrices proving as a fast and reliable alternative for the detection of microorganisms. A power supply of United States standard with 100 - 240 V, 50/60 Hz enables efficient functioning of the reader.

Application

Milliflex® Quantum Power supply- US is an additional accessory for Milliflex® Quantum Rapid Detection System. It finds application in bioburden testing and water monitoring of beer, bottled water, cosmetics, and Pharmaceutical Products Analysis

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service