Skip to Content
Merck
All Photos(1)

Key Documents

MABF2003

Sigma-Aldrich

Anti-CD11a Antibody, clone BU17

clone BU17, from mouse

Synonym(s):

Integrin alpha-L, CD11 antigen-like family member A, Leukocyte adhesion glycoprotein LFA-1 alpha chain, LFA-1A, Leukocyte function-associated molecule 1 alpha chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Pricing and availability is not currently available.

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BU17, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

Related Categories

General description

Integrin alpha-L (UniProt: P20701; also known as CD11 antigen-like family member A, Leukocyte adhesion glycoprotein LFA-1 alpha chain, LFA-1A, Leukocyte function-associated molecule 1 alpha chain, CD11a) is encoded by the ITGAL (also known as CD11A) gene (Gene ID: 3683) in human. Integrins are part of a family of heterodimeric transmembrane glycoproteins that recognize a variety of ligands and control numerous physiologic functions in cells. CD11a is a single-pass type I membrane protein that noncovalently associates with beta2 integrin to forms a heterodimer and serve as a receptor for ICAM1, ICAM2, ICAM3 and ICAM4. It is expressed on B and T lymphocytes, monocytes, macrophages, neutrophils, basophils, and eosinophils. It is also involved in generation of common lymphoid progenitor cells in bone marrow. In resting T-cells, about 40% of CD11a is constitutively phosphorylated and phosphorylation causes conformational changes needed for ligand binding and is considered to be essential for activation by some physiological agents. CD11a is synthesized with a signal peptide (aa 1-25), which is subsequently cleaved off in the mature form. It has a large extracellular domain (aa 26-1090), a transmembrane domain (aa 1091-1111), and a cytoplasmic domain 1112-1170). Three isoforms of CD11a have been described that are produced by alternative splicing.

Specificity

Clone BU17 detects CD11a in human cells. It targets an epitope with in the extracellular domain.

Immunogen

Epitope: extracellular domain
Waldenstrom macroglobulinemia lymphoplasmacytic cells.

Application

Anti-CD11a, clone BU17, Cat. No. MABF2003, is a mouse monoclonal antibody that detects CD11a and has been tested for use in Flow Cytometry.
Flow Cytometry Analysis: A representative lot detected CD11a in Flow Cytometry applications (Baker, M.P., et. al. (1998). Blood. 92(8):2830-43).
Research Category
Inflammation & Immunology

Quality

Evaluated by Flow Cytometry in one million human peripheral blood mononuclear cells.

Flow Cytometry Analysis: 1 µg of this antibody detected CD11a in one million human peripheral blood mononuclear cells.

Target description

128.77 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service